    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CHCHD6 Antibodies
          Proteintech
          CHCHD6 Monoclonal Antibody (2A11E9), CoraLite® 594
          View all (10) CHCHD6 antibodies
          Tech Support
          Cite CHCHD6 Monoclonal Antibody (2A11E9), CoraLite® 594
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          CHCHD6 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CHCHD6 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          CHCHD6 Antibody (CL594-66597) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed MCF-7 cells using CoraLite®594 CHCHD6 antibody (CL594-66597, Clone: 2A11E9) at dilution of 1:400. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CHCHD6 Antibody in Immunocytochemistry (ICC/IF)
          CHCHD6 Antibody in Flow Cytometry (Flow)
          CHCHD6 Antibody in Flow Cytometry (Flow)
          Product Details
          CL594-66597
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:200-1:800
          -
          Flow Cytometry (Flow)
          0.40 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          2A11E9
          Immunogen
          CHCHD6 fusion protein Ag14700
          Conjugate
          CoraLite® 594 CoraLite® 594 CoraLite® 594
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          593/614 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MGSTESSEG RRVSFGVDEE ERVRVLQGVR LSENVVNRMK EPSSPPPAPT SSTFGLQDGN LRAPHKESTL PRSGSSGGQQ PSGMKEGVKR YEQEHAAIQD KLFQVAKRER EAATKHSKAS LPTGEGSISH EEQKSVRLAR ELESREAELR RRDTFYKEQL ERIERKNAEM YKLSSEQFHE AASKMESTIK PRRVEPVCSG LQAQILHCYR DRPHEVLLCS DLVKAYQRCV SAAHKG
          Target Information
          CHCHD6 is a member of a family of proteins containing a conserved (coiled coil 1)-(helix 1)-(coiled coil 2)-(helix 2) domain and has been observed in a complex with the mitochondrial proteins mitofilin, SAM50, metaxins 1 and 2, and CHCHD3. CHCHD6 knockdown causes severe defects in mitochondrial cristae morphology as well as reductions in cell growth, ATP production and oxygen consumption. This decrease of CHCHD6 also led to a reduction in mitofilin protein levels, while a knockdown of mitofilin resulted in a reduced level of CHCHD6, suggesting coordinate regulation between these two proteins. CHCHD6 knockdown in human cancer cells enhances their sensitivity to genotoxic anticancer drugs, suggesting that CHCHD6 may be a potential therapeutic anti-tumor target.
          Bioinformatics
          Protein Aliases: coiled coil helix cristae morphology 1; Coiled-coil-helix cristae morphology protein 1; Coiled-coil-helix-coiled-coil-helix domain-containing protein 6; coiled-coil-helix-coiled-coil-helix domain-containing protein 6, mitochondrial; MICOS complex subunit MIC25; mitochondrial contact site and cristae organizing system subunit 25; protein phosphatase 1, regulatory subunit 23
          View more View less
          Gene Aliases: CHCHD6; CHCM1; MIC25; MICOS25; PPP1R23
          View more View less
          UniProt ID: (Human) Q9BRQ6
          View more View less
          Entrez Gene ID: (Human) 84303
          View more View less
          Function(s)
          protein binding
          Process(es)
          cellular response to DNA damage stimulus inner mitochondrial membrane organization cristae formation
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline