    • See all product categories
  • Applications
    • Cell Analysis
    • Protein Biology
    • Lab Informatics
    • 360° CDMO and CRO Solutions
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
          • Primary Antibodies ›
          • Palladin Antibodies
          Proteintech
          PALLD, palladin Monoclonal Antibody (2A1F6), CoraLite® 594
          View all (13) Palladin antibodies
          Tech Support
          Cite PALLD, palladin Monoclonal Antibody (2A1F6), CoraLite® 594
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          PALLD, palladin Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          Group 53 Created with Sketch.
          PALLD, palladin Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          PALLD, palladin Antibody (CL594-66601) in IHC (PFA)
          Immunofluorescent analysis of (4% PFA) fixed mouse testis tissue using CoraLite®594 PALLD, palladin antibody (CL594-66601, Clone: 2A1F6) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          PALLD, palladin Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          Product Details
          CL594-66601
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Paraffin) (IHC (P))
          1:50-1:500
          -
          Immunohistochemistry (PFA fixed) (IHC (PFA))
          1:200
          -
          Product Specifications
          Species Reactivity
          Human, Pig, Rat
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          2A1F6
          Immunogen
          PALLD,palladin fusion protein Ag1288
          Conjugate
          CoraLite® 594 CoraLite® 594 CoraLite® 594
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          593/614 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: APFFEMKLK HYKIFEGMPV TFTCRVAGNP KPKIYWFKDG KQISPKSDHY TIQRDLDGTC SLHTTASTLD DDGNYTIMAA NPQGRISCTG RLMVQAVNQR GRSPRSPSGH PHVRRPRSRS RDSGDENEPI QERFFRPHFL QAPGDLTVQE GKLCRMDCKV SGLPTPDLSW QLDGKPVRPD SAHKMLVREN GVHSLIIEPV TSRDAGIYTC IATNRAGQNS FSLELVVAAK EAHKPPVFIE KLQNTGVADG YPVRLECRVL GVPPPQIFWK KENESLTHST DRVSMHQDNH GYICLLIQGA TKEDAGWYTV SAKNEAGIVS CTARLDVYTQ WHQQSQSTKP KKVRPSASRY AALSDQGLDI KAAFQPEANP SHLTLNTALV ESEDL
          Target Information
          Palladin is a cytoskeletal protein required for organization of normal actin cytoskeleton. Palladin plays roles in establishing cell morphology, motility, cell adhesion and cell-extracellular matrix interactions in a variety of cell types. Palladin may function as a scaffolding molecule with the potential to impact both actin polymerization as well as the assembly of existing actin filaments into higher-order arrays. It binds to proteins that bind to either monomeric or filamentous actin. It localizes at sites where active actin remodeling takes place, such as lamellipodia and membrane ruffles. Palladin's different isoforms may have functional differences. It is also involved in the control of morphological and cytoskeletal changes associated with dendritic cell maturation.
          Bioinformatics
          Protein Aliases: FLJ22190; FLJ38193; FLJ39139; myoneurin; Palladin; palladin pseudogene; Sarcoma antigen NY-SAR-77; SIH002; unnamed protein product
          View more View less
          Gene Aliases: CGI-151; CGI151; KIAA0992; MYN; PALLD; PNCA1; SIH002
          View more View less
          UniProt ID: (Human) Q8WX93
          View more View less
          Entrez Gene ID: (Pig) 100627292, (Human) 23022, (Rat) 364558
          View more View less
          Function(s)
          actin binding protein binding muscle alpha-actinin binding protein binding involved in cell-cell adhesion scaffold/adaptor protein
          Process(es)
          keratinocyte development epithelial cell morphogenesis cytoskeleton organization homophilic cell adhesion via plasma membrane adhesion molecules axon guidance cell migration actin cytoskeleton organization dendrite self-avoidance
          • Trademarks
          • Fair Trade Fair Trade
          Korea flag icon
          Korea
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910
          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783
          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)
          ISMS Logo
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline