    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
          • Order Status
          • 快速訂購
          • 登入
            登入
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • MTHFR Antibodies
          Proteintech
          MTHFR Monoclonal Antibody (1F7A3), CoraLite® 594
          View all (17) MTHFR antibodies
          Tech Support
          Cite MTHFR Monoclonal Antibody (1F7A3), CoraLite® 594
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          MTHFR Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          MTHFR Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          MTHFR Antibody (CL594-66612) in ICC/IF
          Immunofluorescent analysis of (-20°C Methanol) fixed HeLa cells using CoraLite®594 MTHFR antibody (CL594-66612, Clone: 1F7A3) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          MTHFR Antibody in Immunocytochemistry (ICC/IF)
          MTHFR Monoclonal Antibody (1F7A3), CoraLite® 594
          Product Details
          CL594-66612
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2b
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1F7A3
          Immunogen
          MTHFR fusion protein Ag14970
          3D Epitope / Immunogen
          Conjugate
          CoraLite® 594 CoraLite® 594 CoraLite® 594
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          593/614 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: QELLASGLVP GLHFYTLNRE MATTEVLKRL GMWTEDPRRP LPWALSAHPK RREEDVRPIF WASRPKSYIY RTQEWDEFPN GRWGNSSSPA FGELKDYYLF YLKSKSPKEE LLKMWGEELT SEASVFEVFV LYLSGEPNRN GHKVTCLPWN DEPLAAETSL LKEELLRVNR QGILTINSQP NINGKPSSDP IVGWGPSGGY VFQKAYLEFF TSRETAEALL QVLKKYELRV NYHLVNVKGE NITNAPELQP NAVTWGIFPG REIIQPTVVD PVSFMFWKDE AFALWIEQWG KLYEEESPSR TIIQYIHDNY FLVNLVDNDF PLDNCLWQVV EDTLELLNRP TQNARETEAP
          Target Information
          The protein encoded by this gene catalyzes the conversion of 5,10-methylenetetrahydrofolate to 5-methyltetrahydrofolate, a co-substrate for homocysteine remethylation to methionine. Genetic variation in this gene influences susceptibility to occlusive vascular disease, neural tube defects, colon cancer and acute leukemia, and mutations in this gene are associated with methylenetetrahydrofolate reductase deficiency.
          Bioinformatics
          Protein Aliases: 5,10-methylenetetrahydrofolate reductase (NADPH); methylenetetrahydrofolate reductase (NAD(P)H); Methylenetetrahydrofolate reductase (NADPH); MTHFR; MTHFR long isoform; MTHFR short isoform; RP11-56N19.4; unnamed protein product
          View more View less
          Gene Aliases: MTHFR
          View more View less
          UniProt ID: (Human) P42898
          View more View less
          Entrez Gene ID: (Human) 4524
          View more View less
          Function(s)
          catalytic activity methylenetetrahydrofolate reductase (NAD(P)H) activity protein binding oxidoreductase activity macromolecular complex binding flavin adenine dinucleotide binding NADP binding FAD binding modified amino acid binding methylenetetrahydrofolate reductase (NADPH) activity
          Process(es)
          response to hypoxia neural tube closure methionine metabolic process methionine biosynthetic process response to xenobiotic stimulus response to vitamin B2 tetrahydrofolate interconversion response to amino acid S-adenosylmethionine metabolic process tetrahydrofolate metabolic process homocysteine metabolic process response to folic acid response to interleukin-1 heterochromatin organization
          Hong Kong flag icon
          Hong Kong
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline