    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • STRAP Antibodies
          Proteintech
          STRAP Monoclonal Antibody (5E11F3), CoraLite® 594
          View all (14) STRAP antibodies
          Tech Support
          Cite STRAP Monoclonal Antibody (5E11F3), CoraLite® 594
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          STRAP Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          STRAP Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          STRAP Antibody (CL594-66712) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed HEK-293 cells using CoraLite®594 STRAP antibody (CL594-66712, Clone: 5E11F3) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          STRAP Antibody in Immunocytochemistry (ICC/IF)
          Product Details
          CL594-66712
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          5E11F3
          Immunogen
          STRAP fusion protein Ag13085
          Conjugate
          CoraLite® 594 CoraLite® 594 CoraLite® 594
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          593/614 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MAMRQTPLT CSGHTRPVVD LAFSGITPYG YFLISACKDG KPMLRQGDTG DWIGTFLGHK GAVWGATLNK DATKAATAAA DFTAKVWDAV SGDELMTLAH KHIVKTVDFT QDSNYLLTGG QDKLLRIYDL NKPEAEPKEI SGHTSGIKKA LWCSEDKQIL SADDKTVRLW DHATMTEVKS LNFNMSVSSM EYIPEGEILV ITYGRSIAFH SAVSLDPIKS FEAPATINSA SLHPEKEFLV AGGEDFKLYK YDYNSGEELE SYKGHFGPIH CVRFSPDGEL YASGSEDGTL RLWQTVVGKT YGLWKCVLPE EDSGELAKPK IGFPETTEEE LEEIASENSD CIFPSAPDVK A
          Target Information
          STRAP is a 350 amino acid MAP activator with WD-40 repeats that is localized both in the cytoplasm and the nucleus. It is a ubiquitously expressed protein that acts as an inhibitor of TGF-beta signaling and an important regulator of cell proliferation. Stable expression of STRAP results in activation of the mitogen-activated protein kinase/extracellular signal-regulated kinase pathway and in down-regulation of the cyclin-dependent kinase inhibitor p21 (Cip1), which results in retinoblastoma protein hyperphosphorylation. Upregulation of STRAP might play a role in tumor development as an early event for colorectal cancers. It is the first component of the U snRNP assembly machinery that associates with SMN complex in a compartment-specific way and plays a crucial role in the intracellular distribution of SMN1.
          Bioinformatics
          Protein Aliases: MAP activator with WD repeats; MAPK activator with WD repeats; mitogen-activated protein kinase activator with WD repeats; Serine-threonine kinase receptor-associated protein; unnamed protein product; UNR-interacting protein; WD-40 repeat protein PT-WD
          View more View less
          Gene Aliases: AW557906; C78091; C79202; MAWD; PT-WD; STRAP; UNRIP
          View more View less
          UniProt ID: (Human) Q9Y3F4, (Mouse) Q9Z1Z2, (Rat) Q5XIG8
          View more View less
          Entrez Gene ID: (Human) 11171, (Mouse) 20901, (Rat) 297699
          View more View less
          Function(s)
          RNA binding mRNA binding receptor binding protein binding U2 snRNP binding poly(A) RNA binding translation initiation factor
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter alternative mRNA splicing, via spliceosome spliceosomal snRNP assembly mRNA splicing, via spliceosome mRNA processing RNA splicing ribonucleoprotein complex assembly neuron differentiation maintenance of gastrointestinal epithelium negative regulation of transforming growth factor beta receptor signaling pathway negative regulation of epithelial cell migration negative regulation of epithelial to mesenchymal transition negative regulation of epithelial cell proliferation negative regulation of pathway-restricted SMAD protein phosphorylation
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline