    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
  • Services
    • 360° CDMO and CRO Solutions
          • Primary Antibodies ›
          • SBDS Antibodies
          Proteintech
          SBDS Monoclonal Antibody (1F8A10), CoraLite® 594
          View all (12) SBDS antibodies
          Tech Support
          Cite SBDS Monoclonal Antibody (1F8A10), CoraLite® 594
          Additional Information:
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          SBDS Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SBDS Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          SBDS Antibody (CL594-67200) in WB
          Various lysates were subjected to SDS PAGE followed by western blot with CL594-67200 (SBDS antibody) at dilution of 1:1,000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          SBDS Antibody in Western Blot (WB)
          SBDS Antibody in Flow Cytometry (Flow)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          Product Details
          CL594-67200
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:2,000
          -
          Flow Cytometry (Flow)
          1x10^6 cells
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1F8A10
          Immunogen
          SBDS fusion protein Ag11731
          Conjugate
          CoraLite® 594 CoraLite® 594 CoraLite® 594
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          593/614 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MSIFTPTNQ IRLTNVAVVR MKRAGKRFEI ACYKNKVVGW RSGVEKDLDE VLQTHSVFVN VSKGQVAKKE DLISAFGTDD QTEICKQILT KGEVQVSDKE RHTQLEQMFR DIATIVADKC VNPETKRPYT VILIERAMKD IHYSVKTNKS TKQQALEVIK QLKEKMKIER AHMRLRFILP VNEGKKLKEK LKPLIKVIES EDYGQQLEIV CLIDPGCFRE IDELIKKETK GKGSLEVLNL KDVEEGDEKF E
          Target Information
          This gene encodes a member of a highly conserved protein family that exists from archaea to vertebrates and plants. The encoded protein may function in RNA metabolism. Mutations within this gene are associated with Shwachman-Bodian-Diamond syndrome. An alternative transcript has been described, but its biological nature has not been determined. This gene has a closely linked pseudogene that is distally located.
          Bioinformatics
          Protein Aliases: FLJ10917; protein 22A3; SBDS; SBDS ribosome assembly guanine nucleotide exchange factor; SBDS, ribosome assembly guanine nucleotide exchange factor; Shwachman-Bodian-Diamond syndrome homolog; shwachman-Bodian-Diamond syndrome protein homolog; unnamed protein product
          View more View less
          Gene Aliases: 4733401P19Rik; CGI-97; SDO1; SDS; SWDS
          View more View less
          UniProt ID: (Human) Q9Y3A5, (Mouse) P70122, (Rat) Q5RK30
          View more View less
          Entrez Gene ID: (Human) 51119, (Mouse) 66711, (Rat) 288615
          View more View less
          Function(s)
          RNA binding protein binding microtubule binding rRNA binding ribosome binding
          Process(es)
          hematopoietic progenitor cell differentiation rRNA processing mitotic spindle organization bone mineralization leukocyte chemotaxis ribosome biogenesis mature ribosome assembly bone marrow development inner cell mass cell proliferation biological_process
          Hong Kong flag icon
          Hong Kong
          Your items have has been added!
          Host server : magellan-search-blue-74d7b4b4d9-v4tl5:80/100.66.78.131:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline