    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
  • Services
    • 360° CDMO and CRO Solutions
          • Primary Antibodies ›
          • TH1L Antibodies
          Proteintech
          TH1L Monoclonal Antibody (1A10G2), CoraLite® 594
          View all (8) TH1L antibodies
          Tech Support
          Cite TH1L Monoclonal Antibody (1A10G2), CoraLite® 594
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          TH1L Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          TH1L Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          TH1L Antibody (CL594-67380) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed HeLa cells using CoraLite®594 TH1L antibody (CL594-67380, Clone: 1A10G2) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          TH1L Antibody in Immunocytochemistry (ICC/IF)
          TH1L Antibody in Flow Cytometry (Flow)
          Product Details
          CL594-67380
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Flow Cytometry (Flow)
          0.80 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1A10G2
          Immunogen
          TH1L fusion protein Ag24894
          Conjugate
          CoraLite® 594 CoraLite® 594 CoraLite® 594
          • Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Excitation/Emission Max
          593/614 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: QAMMSVLAQ EEQGGSAVRR IAQEVQRFAQ EKGHDASQIT LALGTAASYP RACQALGAML SKGALNPADI TVLFKMFTSM DPPPVELIRV PAFLDLFMQS LFKPGARINQ DHKHKYIHIL AYAASVVETW KKNKRVSINK DELKSTSKAV ETVHNLCCNE NKGASELVAE LSTLYQCIRF PVVAMGVLKW VDWTVSEPRY FQLQTDHTPV HLALLDEIST CHQLLHPQVL QLLVKLFETE HSQLDVMEQL ELKKTLLDRM VHLLSRGYVL PVVSYIRKCL EKLDTDISLI RYFVTEVLDV IAPPYTSDFV QLFLPILEND SIAGTIKTEG EHDPVTEFIA HCKSNFIMVN
          Target Information
          The NELF complex of proteins interacts with the DSIF protein complex to repress transcriptional elongation by RNA polymerase II. The protein encoded by this gene is an essential part of the NELF complex. Alternative translation initiation site usage results in the formation of two isoforms with different N-termini.
          Bioinformatics
          Protein Aliases: Negative elongation factor C/D; negative elongation factor complex member C/D,Th1l; Negative elongation factor D; negative elongation factor proteins C and D; NELF-C/D; NELF-D; TH1 drosophila homolog; TH1 homolog; TH1-like homolog; TH1-like protein; trihydrophobin 1; trihydrophobin 1 homolog; unnamed protein product
          View more View less
          Gene Aliases: 2410003I03Rik; C77797; HSPC130; NELF-C; NELF-D; NELFCD; NELFD; TH1; TH1L
          View more View less
          UniProt ID: (Human) Q8IXH7, (Mouse) Q922L6
          View more View less
          Entrez Gene ID: (Human) 51497, (Rat) 679203, (Mouse) 57314
          View more View less
          Function(s)
          RNA binding protein binding molecular_function
          Process(es)
          negative regulation of transcription elongation from RNA polymerase II promoter negative regulation of transcription, DNA-templated biological_process transcription, DNA-templated regulation of transcription, DNA-templated
          Hong Kong flag icon
          Hong Kong
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline