    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • ZG16 Antibodies
          Proteintech
          ZG16 Monoclonal Antibody (1A7B9), CoraLite® Plus 594
          View all (2) ZG16 antibodies
          Tech Support
          Cite ZG16 Monoclonal Antibody (1A7B9), CoraLite® Plus 594
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          ZG16 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          Group 53 Created with Sketch.
          ZG16 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          ZG16 Antibody (CL594-67389) in IHC (PFA)
          Immunofluorescent analysis of (4% PFA) fixed mouse small intestine tissue using CoraLite® Plus 594 ZG16 antibody (CL594-67389, Clone: 1A7B9) at dilution of 1:100, CoraLite® Plus 488 PIGR antibody (CL488-22024, green), Villin antibody (16488-1-AP, Magenta). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          ZG16 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          ZG16 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          ZG16 Monoclonal Antibody (1A7B9), CoraLite® Plus 594
          Product Details
          CL594-67389
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Paraffin) (IHC (P))
          1:50-1:500
          -
          Immunohistochemistry (PFA fixed) (IHC (PFA))
          1:100
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Pig, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1A7B9
          Immunogen
          ZG16 fusion protein Ag11332
          3D Epitope / Immunogen
          Conjugate
          CoraLite® Plus 594 CoraLite® Plus 594 CoraLite® Plus 594
          • Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Excitation/Emission Max
          588/604 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MLTVALLALL CASASGNAIQ ARSSSYSGEY GSGGGKRFSH SGNQLDGPIT ALRVRVNTYY IVGLQVRYGK VWSDYVGGRN GDLEEIFLHP GESVIQVSGK YKWYLKKLVF VTDKGRYLSF GKDSGTSFNA VPLHPNTVLR FISGRSGSLI DAIGLHWDVY PTSCSRC
          Bioinformatics
          Protein Aliases: jacalin-like lectin domain containing; Secretory lectin ZG16; unnamed protein product; Zymogen granule membrane protein 16; Zymogen granule protein 16; zymogen granule protein 16 homolog
          View more View less
          Gene Aliases: 1810010M01Rik; AI593689; JCLN; JCLN1; Zg-16p; ZG16; ZG16A; ZG16p
          View more View less
          UniProt ID: (Human) O60844, (Mouse) Q8K0C5, (Rat) Q8CJD3
          View more View less
          Entrez Gene ID: (Pig) 100620560, (Human) 653808, (Mouse) 69036, (Rat) 171449
          View more View less
          Function(s)
          protein binding carbohydrate binding peptidoglycan binding molecular_function
          Process(es)
          transport biological_process protein transport defense response to Gram-positive bacterium suppression of symbiont entry into host
          Argentina flag icon
          Argentina
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline