    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • PPP4R1 Antibodies
          Proteintech
          PPP4R1 Monoclonal Antibody (3D12G12), CoraLite® 594
          View all (9) PPP4R1 antibodies
          Tech Support
          Cite PPP4R1 Monoclonal Antibody (3D12G12), CoraLite® 594
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          PPP4R1 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          Group 53 Created with Sketch.
          PPP4R1 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          PPP4R1 Antibody (CL594-67392) in IHC (PFA)
          Immunofluorescent analysis of (4% PFA) fixed mouse liver tissue using CoraLite®594 PPP4R1 antibody (CL594-67392, Clone: 3D12G12) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          PPP4R1 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          PPP4R1 Antibody in Flow Cytometry (Flow)
          Product Details
          CL594-67392
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Paraffin) (IHC (P))
          1:50-1:500
          -
          Immunohistochemistry (PFA fixed) (IHC (PFA))
          1:200
          -
          Flow Cytometry (Flow)
          0.40 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          3D12G12
          Immunogen
          PPP4R1 fusion protein Ag26750
          Conjugate
          CoraLite® 594 CoraLite® 594 CoraLite® 594
          • Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Excitation/Emission Max
          593/614 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: NRTRDQEAP EDVQVRPEDT PSDLSVSNSS VILENTMEDH AAEASGKPLG EISVPLDSSL LCTLSSESHQ EAASNENDKK PGNYKSMLRP EVGTTSQDSA LLDQELYNSF HFWRTPLPEI DLDIELEQNS GGKPSPEGPE EESEGPVPSS PNITMATRKE LEEMIENLEP HIDDPDVKAQ VEVLSAALRA S
          Target Information
          Regulatory subunit of serine/threonine-protein phosphatase 4. May play a role in regulation of cell division in renal glomeruli. The PPP4C-PPP4R1 PP4 complex may play a role in dephosphorylation and regulation of HDAC3.
          Bioinformatics
          Protein Aliases: meg-1; serine/threonine phosphatase 4; Serine/threonine-protein phosphatase 4 regulatory subunit 1; splice variant induced by elevated nuclear concentration of splicing factor SLU7; unnamed protein product
          View more View less
          Gene Aliases: 3110001J10Rik; MEG1; PP4(Rmeg); PP4R1; PPP4R1
          View more View less
          UniProt ID: (Human) Q8TF05, (Mouse) Q8K2V1
          View more View less
          Entrez Gene ID: (Human) 9989, (Mouse) 70351
          View more View less
          Function(s)
          phosphoprotein phosphatase activity protein binding protein phosphatase regulator activity molecular_function phosphatase modulator
          Process(es)
          protein phosphorylation signal transduction biological_process
          • News News
          • Responsibility Responsibility
          Chile flag icon
          Chile
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline