    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • UAP1 Antibodies
          Proteintech
          UAP1 Monoclonal Antibody (2C7B2), CoraLite® 594
          View all (24) UAP1 antibodies
          Tech Support
          Cite UAP1 Monoclonal Antibody (2C7B2), CoraLite® 594
          Additional Information:
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          UAP1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          UAP1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          UAP1 Antibody (CL594-67545) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed PC-3 cells using CoraLite®594 UAP1 antibody (CL594-67545, Clone: 2C7B2) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          UAP1 Antibody in Immunocytochemistry (ICC/IF)
          UAP1 Antibody in Flow Cytometry (Flow)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          Product Details
          CL594-67545
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Flow Cytometry (Flow)
          0.40 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          2C7B2
          Immunogen
          UAP1 fusion protein Ag9513
          Conjugate
          CoraLite® 594 CoraLite® 594 CoraLite® 594
          • Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Excitation/Emission Max
          593/614 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MNINDLKLT LSKAGQEHLL RFWNELEEAQ QVELYAELQA MNFEELNFFF QKAIEGFNQS SYQKNVDARM EPVPREVLGS ATRDQDQLQA WESEGLFQIS QNKVAVLLLA GGQGTRLGVA YPKGMYDVGL PSRKTLFQIQ AERILKLQQV AEKYYGNKCI IPWYIMTSGR TMESTKEFFT KHKYFGLKKE NVIFFQQGML PAMSFDGKII LEEKNKVSMA PDGNGGLYRA LAAQNIVEDM EQRGIWSIHV YCVDNILVKV ADPRFIGFCI QKGADCGAKV VEKTNPTEPV GVVCRVDGVY QVVEYSEISL ATAQKRSSDG RLLFNAGNIA NHFFTVPFLR DVVNVYEPQL QHH
          Target Information
          Converts UDP and GlcNAc-1-P into UDP-GlcNAc, and UDP and GalNAc-1-P into UDP-GalNAc. Isoform AGX1 has 2 to 3 times higher activity towards GalNAc-1-P, while isoform AGX2 has 8 times more activity towards GlcNAc-1-P.
          Bioinformatics
          Protein Aliases: antigen X; protein-pyrophosphorylation enzyme; sperm associated antigen 2; testis tissue sperm-binding protein Li 37a; UDP-N-acetylgalactosamine pyrophosphorylase; UDP-N-acetylglucosamine diphosphorylase 1; UDP-N-acetylhexosamine pyrophosphorylase 1; unnamed protein product
          View more View less
          Gene Aliases: AGX; AGX1; AGX2; SPAG2
          View more View less
          UniProt ID: (Human) Q16222
          View more View less
          Entrez Gene ID: (Human) 6675
          View more View less
          Function(s)
          UDP-N-acetylglucosamine diphosphorylase activity transferase activity, transferring phosphorus-containing groups identical protein binding UDP-N-acetylgalactosamine diphosphorylase activity uridylyltransferase activity protein serine pyrophosphorylase activity
          Process(es)
          UDP-N-acetylglucosamine biosynthetic process positive regulation of type I interferon production antiviral innate immune response
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-green-79565875f6-vjw2z:80/100.66.77.38:80.
          git-commit: 280dcbe45276d5d6e613c14d399020b8a49b7116
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.57.0-Offline