    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • RPL6 Antibodies
          Proteintech
          RPL6 Monoclonal Antibody (3F6F8), CoraLite® 594
          View all (9) RPL6 antibodies
          Tech Support
          Cite RPL6 Monoclonal Antibody (3F6F8), CoraLite® 594
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          RPL6 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          RPL6 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          RPL6 Antibody (CL594-67729) in ICC/IF
          Immunofluorescent analysis of (-20°C Ethanol) fixed HepG2 cells using CoraLite®594 RPL6 antibody (CL594-67729, Clone: 3F6F8) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          RPL6 Antibody in Immunocytochemistry (ICC/IF)
          Product Details
          CL594-67729
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          3F6F8
          Immunogen
          RPL6 fusion protein Ag6840
          Conjugate
          CoraLite® 594 CoraLite® 594 CoraLite® 594
          • Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Excitation/Emission Max
          593/614 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MAGEKVEKP DTKEKKPEAK KVDAGGKVKK GNLKAKKPKK GKPHCSRNPV LVRGIGRYSR SAMYSRKAMY KRKYSAAKSK VEKKKKEKVL ATVTKPVGGD KNGGTRVVKL RKMPRYYPTE DVPRKLLSHG KKPFSQHVRK LRASITPGTI LIILTGRHRG KRVVFLKQLA SGLLLVTGPL VLNRVPLRRT HQKFVIATST KIDISNVKIP KHLTDAYFKK KKLRKPRHQE GEIFDTEKEK YEITEQRKID QKAVDSQILP KIKAIPQLQG YLRSVFALTN GIYPHKLVF
          Target Information
          Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L6E family of ribosomal proteins. It is located in the cytoplasm. The protein can bind specifically to domain C of the tax-responsive enhancer element of human T-cell leukemia virus type 1, and it has been suggested that the protein may participate in tax-mediated transactivation of transcription. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. Two alternatively spliced transcript variants encoding the same protein have been found for this gene.
          Bioinformatics
          Protein Aliases: 60S ribosomal protein L6; DNA-binding protein TAXREB107; Large ribosomal subunit protein eL6; Neoplasm-related protein C140; TAX-responsive enhancer element-binding protein 107; TAXREB107; unnamed protein product
          View more View less
          Gene Aliases: eL6; L6; RPL6; SHUJUN-2; TAXREB107; TXREB1
          View more View less
          UniProt ID: (Human) Q02878, (Mouse) P47911, (Rat) P21533
          View more View less
          Entrez Gene ID: (Human) 6128, (Mouse) 19988, (Rat) 117042
          View more View less
          Function(s)
          DNA binding RNA binding structural constituent of ribosome protein binding cadherin binding poly(A) RNA binding ribosomal protein
          Process(es)
          cytoplasmic translation regulation of transcription, DNA-templated translation
          • News News
          • Responsibility Responsibility
          Chile flag icon
          Chile
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline