          • Primary Antibodies ›
          • RAB1B Antibodies
          Proteintech
          RAB1B Monoclonal Antibody (3C2G6), CoraLite® 594
          View all (7) RAB1B antibodies
          Tech Support
          Cite RAB1B Monoclonal Antibody (3C2G6), CoraLite® 594
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          RAB1B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          RAB1B Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          RAB1B Antibody (CL594-67837) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed NIH/3T3 cells using CoraLite®594 RAB1B antibody (CL594-67837, Clone: 3C2G6) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          RAB1B Antibody in Immunocytochemistry (ICC/IF)
          RAB1B Antibody in Flow Cytometry (Flow)
          Product Details
          CL594-67837
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Flow Cytometry (Flow)
          0.40 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          3C2G6
          Immunogen
          RAB1B fusion protein Ag12069
          Conjugate
          CoraLite® 594 CoraLite® 594 CoraLite® 594
          • Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Excitation/Emission Max
          593/614 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MNPEYDYLF KLLLIGDSGV GKSCLLLRFA DDTYTESYIS TIGVDFKIRT IELDGKTIKL QIWDTAGQER FRTITSSYYR GAHGIIVVYD VTDQESYANV KQWLQEIDRY ASENVNKLLV GNKSDLTTKK VVDNTTAKEF ADSLGIPFLE TSAKNATNVE QAFMTMAAEI KKRMGPGAAS GGERPNLKID STPVKPAGGG CC
          Target Information
          The small GTPases Rab are key regulators of intracellular membrane trafficking, from the formation of transport vesicles to their fusion with membranes. Rabs cycle between an inactive GDP-bound form and an active GTP-bound form that is able to recruit to membranes different set of downstream effectors directly responsible for vesicle formation, movement, tethering and fusion. RAB1B regulates vesicular transport between the endoplasmic reticulum and successive Golgi compartments. Plays a role in the initial events of the autophagic vacuole development which take place at specialized regions of the endoplasmic reticulum.
          Bioinformatics
          Protein Aliases: RAB1B; RAB1B-like, member RAS oncogene family; Ras-related protein Rab-1B; small GTP-binding protein; unnamed protein product
          View more View less
          Gene Aliases: 1110011F09Rik; RAB1B; Rab1bl
          View more View less
          UniProt ID: (Human) Q9H0U4, (Mouse) Q9D1G1
          View more View less
          Entrez Gene ID: (Human) 81876, (Rat) 100126191, (Mouse) 76308
          View more View less
          Function(s)
          nucleotide binding GTPase activity obsolete small monomeric GTPase activity protein binding GTP binding hydrolase activity GDP binding
          Process(es)
          autophagosome assembly intracellular protein transport ER to Golgi vesicle-mediated transport autophagy Golgi organization protein transport virion assembly establishment of endothelial intestinal barrier positive regulation of glycoprotein metabolic process regulation of autophagosome assembly metabolic process Rab protein signal transduction transport small GTPase mediated signal transduction
          Pedidos Plus Icon Minus Icon
          • Status do pedido
          • Ajuda para pedidos
          • Pedido rápido
          • Supply Center
          • eProcurement
          Suporte Plus Icon Minus Icon
          • Ajuda e suporte
          • Entre em Contato
          • Centros de Suporte Técnico
          • Obter Documentos e Certificados
          • Informe um Problema no Site
          Recursos Plus Icon Minus Icon
          • Centros de aprendizagem
          • Promoções
          • Eventos & Webinars
          • Mídia Sociais
          Sobre a Thermo Fisher Plus Icon Minus Icon
          • Sobre Nós Sobre Nós
          • Carreiras Carreiras
          • Investidores Investidores
          • Sala de Impresa Sala de Impresa
          • Responsabilidade Social Responsabilidade Social
          • Marcas
          • Políticas e avisos
          Nosso Portfólio Plus Icon Minus Icon
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline