    • See all product categories
  • Applications
    • Cell Analysis
    • Protein Biology
    • Lab Informatics
    • 360° CDMO and CRO Solutions
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
          • Primary Antibodies ›
          • TNIK Antibodies
          Proteintech
          TNIK Monoclonal Antibody (1E9H3), CoraLite® 594
          View all (20) TNIK antibodies
          Tech Support
          Cite TNIK Monoclonal Antibody (1E9H3), CoraLite® 594
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          TNIK Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          TNIK Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          TNIK Antibody (CL594-67948) in ICC/IF
          Immunofluorescent analysis of (4% PFA) fixed U2OS cells using CoraLite®594 TNIK antibody (CL594-67948, Clone: 1E9H3) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          TNIK Antibody in Immunocytochemistry (ICC/IF)
          TNIK Antibody in Flow Cytometry (Flow)
          Product Details
          CL594-67948
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:500
          -
          Flow Cytometry (Flow)
          0.80 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1E9H3
          Immunogen
          TNIK fusion protein Ag30710
          Conjugate
          CoraLite® 594 CoraLite® 594 CoraLite® 594
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          593/614 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: PRVYQTSPT DEDEEDEESS ATALFTSELL RQEQAKLNEA RKISVVNVNP TNIRPHSDTP EIRKYKKRFN SEILCAALWG VNLLVGTENG LMLLDRSGQG KVYNLINRRR FQQMDVLEGL NVLVTISGKK NKLRVYYLSW LRNRILHNDP EVEKKQGWIT VGDLEGCIHY KVVKYERIKF LVIALKNAVE IYAWAPKPYH KFMAFKSFAD LQHKPLLVDL TVEEGQRLKV IFGSHTGFHV IDVDSGNSYD IYIPSHIQGN ITPHAIVILP KTDGMEMLVC YEDEGVYVNT YGRITKDVVL QWGEMPTSVA YIHSNQIMGW GEKAIEIRSV ETGHLDGVFM HKRAQRLKFL CERNDKVFFA SVRSGGSSQV FFMTLNRNSM MNW
          Target Information
          TNIK or TRAF2 and NCK interacting kinase is characterized by an N-terminal kinase domain and a C-terminal GCK domain that serves a regulatory function. TNIK is mainly expression in brain, heart, and spleen and it is a specific effector of RAP2 which regulate actin cytoskeleton. TNIK is autophosphorylated in a manner dependent upon lys54 in the ATP-binding pocket of its kinase domain and plays a main role in cytoskeleton regulation. TNIK has also been shown to phosphorylate gelsolin, the principal intracellular and extracellular actin-severing protein, in vitro. This and evidence from mutational studies suggest that TNIK functions in the regulation of the cytoskeleton.
          Bioinformatics
          Protein Aliases: TRAF2 and NCK-interacting protein kinase; unnamed protein product
          View more View less
          Gene Aliases: KIAA0551; MRT54; TNIK
          View more View less
          UniProt ID: (Human) Q9UKE5
          View more View less
          Entrez Gene ID: (Human) 23043
          View more View less
          Function(s)
          nucleotide binding protein kinase activity protein serine/threonine kinase activity protein binding ATP binding kinase activity transferase activity protein serine kinase activity
          Process(es)
          MAPK cascade protein phosphorylation cytoskeleton organization nervous system development Wnt signaling pathway actin cytoskeleton organization intracellular signal transduction regulation of MAPK cascade regulation of JNK cascade positive regulation of JNK cascade protein autophosphorylation neuron projection morphogenesis regulation of dendrite morphogenesis positive regulation of intracellular signal transduction positive regulation of microvillus assembly
          • Trademarks
          • Fair Trade Fair Trade
          Korea flag icon
          Korea
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910
          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783
          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)
          ISMS Logo
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline