  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • Albumin Antibodies
          Proteintech
          Albumin Recombinant Rabbit Monoclonal Antibody (4K1), CoraLite® 594
          View all (42) Albumin antibodies
          Tech Support
          Cite Albumin Recombinant Rabbit Monoclonal Antibody (4K1), CoraLite® 594
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          Albumin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Albumin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Albumin Antibody (CL594-82783-6) in IHC (P)
          Immunofluorescent analysis of (4% PFA) fixed paraffin-embedded mouse liver tissue using CoraLite594 Albumin antibody (CL594-82783-6, Clone: 4K1 ) at dilution of 1:200. Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Albumin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Albumin Recombinant Rabbit Monoclonal Antibody (4K1), CoraLite® 594
          Product Details
          CL594-82783-6
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Paraffin) (IHC (P))
          1:50-1:500
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          CHO cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          4K1
          Immunogen
          Albumin fusion protein Ag9569
          3D Epitope / Immunogen
          Conjugate
          CoraLite® 594 CoraLite® 594 CoraLite® 594
          • Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Excitation/Emission Max
          593/614 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: SKLVTDLTKV HTECCHGDLL ECADDRADLA KYICENQDSI SSKLKECCEK PLLEKSHCIA EVENDEMPAD LPSLAADFVE SKDVCKNYAE AKDVFLGMFL YEYARRHPDY SVVLLLRLAK TYETTLEKCC AAADPHECYA KVFDEFKPLV EEPQNLIKQN CELFEQLGEY KFQNALLVRY TKKVPQVSTP TLVEVSRNLG KVGSKCCKHP EAKRMPCAED YLSVVLNQLC VLHEKTPVSD RVTKCCTESL VNRRPCFSAL EVDETYVPKE FNAETFTFHA DICTLSEKER QIKKQTALVE LVKHKPKATK EQLKAVMDDF AAFVEKCCKA DDKETCFAEE GKKLVAASQA ALGL
          Target Information
          Albumin is a soluble, monomeric protein which comprises about one-half of the blood serum protein. Albumin functions primarily as a carrier protein for steroids, fatty acids, and thyroid hormones and plays a role in stabilizing extracellular fluid volume. Albumin is a globular unglycosylated serum protein of molecular weight 65,000. Albumin is synthesized in the liver as preproalbumin which has an N-terminal peptide that is removed before the nascent protein is released from the rough endoplasmic reticulum. The product, proalbumin, is in turn cleaved in the Golgi vesicles to produce the secreted albumin.
          Bioinformatics
          Protein Aliases: Albumin; albumin 1; GIG20; GIG42; serum albumin; unnamed protein product
          View more View less
          Gene Aliases: ALB; Alb-1; Alb1; Albza; FDAHT; GIG20; GIG42; HSA; PRO0883; PRO0903; PRO1341; PRO1708; PRO2044; PRO2619; PRO2675; UNQ696/PRO1341
          View more View less
          UniProt ID: (Human) P02768, (Rat) P02770, (Mouse) P07724
          View more View less
          Entrez Gene ID: (Human) 213, (Rat) 24186, (Mouse) 11657
          View more View less
          Function(s)
          DNA binding fatty acid binding copper ion binding protein binding lipid binding toxic substance binding antioxidant activity oxygen binding pyridoxal phosphate binding identical protein binding metal ion binding chaperone binding exogenous protein binding enterobactin binding drug binding zinc ion binding enzyme binding transfer/carrier protein
          Process(es)
          cellular response to starvation response to nutrient levels negative regulation of mitochondrial depolarization cellular response to calcium ion starvation cellular oxidant detoxification retina homeostasis transport response to nutrient response to organic substance hemolysis by symbiont of host erythrocytes vasodilation negative regulation of apoptotic process positive regulation of circadian sleep/wake cycle, non-REM sleep response to mercury ion maintenance of mitochondrion location response to platinum ion
          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline