    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
            • Aspire Member Program
            • Connect: Lab, Data, Apps
          • Primary Antibodies ›
          • TER-119 Antibodies
          Proteintech
          TER-119 (TER-119) Monoclonal Antibody (TER-119), CoraLite® Plus 647
          View all (39) TER-119 antibodies
          Questions & Answers
          Cite TER-119 (TER-119) Monoclonal Antibody (TER-119), CoraLite® Plus 647
          • Antibody Testing Data (1)
          TER-119 (TER-119) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          TER-119 (TER-119) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          TER-119 (TER-119) Antibody (CL647-65149) in Flow
          1x10^6 mouse bone marrow cells were surface co-stained with CoraLite®488 Anti-Mouse CD45 (30-F11) and 0.25 µg CoraLite® Plus 647 Anti-Mouse TER-119 (CL647-65149, Clone: TER-119) or 0.25 µg CoraLite® Plus 647-conjugated Rat IgG2b isotype control. Cells were not fixed. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          TER-119 (TER-119) Antibody in Flow Cytometry (Flow)
          Product Details
          CL647-65149
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Mouse
          Host/Isotype
          Rat / IgG2b, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          TER-119
          Immunogen
          Day-14 fetal liver cells from a C57BL/6 mouse
          Conjugate
          CoraLite® Plus 647 CoraLite® Plus 647 CoraLite® Plus 647
          • Unconjugated
          • APC 
          • CoraLite Plus 488
          • FITC
          • PE 
          • Request custom conjugation
          Excitation/Emission Max
          654/674 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA
          Contains
          0.1% sodium azide
          Storage conditions
          4°C, store in dark
          Shipping conditions
          Wet ice
          Target Information
          TER-119 is a lineage marker for erythroid cells from early proerythroblast to mature erythrocyte stages in adult blood, spleen, and bone marrow. It is also present in yolk sac, and fetal and newborn liver. The TER-119 antigen is not expressed by cells of earlier erythroid development at BFU-e (blast-forming unit erythroid) stage or CFU-e (colony-forming unit erythroid) stage.
          Bioinformatics
          Protein Aliases: Ly-76; Ly76; TER119
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States
          Your items have has been added!
          Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline