    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • eIF3l Antibodies
          Proteintech
          EIF3L Monoclonal Antibody (1B11A10), CoraLite® Plus 647
          View all (8) eIF3l antibodies
          Tech Support
          Cite EIF3L Monoclonal Antibody (1B11A10), CoraLite® Plus 647
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          EIF3L Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          EIF3L Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          EIF3L Antibody (CL647-67719) in Flow
          1x10^6 HeLa cells were intracellularly stained with 0.8 µg CoraLite® Plus 647 Anti-Human EIF3L (CL647-67719, Clone: 1B11A10) (red), or 0.8 µg CoraLite® Plus 647 Mouse IgG2a Isotype Control (C1.18.4) (CL647-65208, Clone: C1.18.4) (blue). Cells were fixed with 4% PFA and permeabilized with Flow Cytometry Perm Buffer (PF00011-C). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          EIF3L Antibody in Flow Cytometry (Flow)
          Product Details
          CL647-67719
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          0.80 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1B11A10
          Immunogen
          EIF3L fusion protein Ag26680
          Conjugate
          CoraLite® Plus 647 CoraLite® Plus 647 CoraLite® Plus 647
          • Unconjugated
          • CoraLite 594
          • CoraLite Plus 488
          • Request custom conjugation
          Excitation/Emission Max
          654/674 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: SVAGEYGRH SLYKMLGYFS LVGLLRLHSL LGDYYQAIKV LENIELNKKS MYSRVPECQV TTYYYVGFAY LMMRRYQDAI RVFANILLYI QRTKSMFQRT TYKYEMINKQ NEQMHALLAI ALTMYPMRID ESIHLQLREK YGDKMLRMQK GDPQVYEELF SYSCPKFLSP VVPNYDNVHP NYHKEPFLQQ LKVFSDEVQQ QAQLSTIRSF LKLYTTMPVA KLAGFLDLTE QEFRIQLLVF KHKMKNLVWT SGISALDGEF QSASEVDFYI DKDMIHIADT KVARRYGDFF IRQIHKFEEL NRTLKKMGQR P
          Target Information
          Component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis (PubMed:17581632, PubMed:25849773, PubMed:27462815). The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S pre-initiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of post-termination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation (PubMed:17581632). The eIF-3 complex specifically targets and initiates translation of a subset of mRNAs involved in cell proliferation, including cell cycling, differentiation and apoptosis, and uses different modes of RNA stem-loop binding to exert either translational activation or repression (PubMed:25849773). [UniProt]
          Bioinformatics
          Protein Aliases: 66 kDa tyrosine-rich heat shock protein; 67 kDa polymerase-associated factor; dJ1014D13.1; eIEF associated protein HSPC021; eIF3l; Eukaryotic translation initiation factor 3 subunit 6-interacting protein; Eukaryotic translation initiation factor 3 subunit E-interacting protein; Eukaryotic translation initiation factor 3 subunit L; eukaryotic translation initiation factor 3, subunit 6 interacting protein; HSP-66Y; PAF67; unnamed protein product
          View more View less
          Gene Aliases: 0610011H21Rik; D15N1e; EIF3EIP; Eif3ip; EIF3L; EIF3S11; EIF3S6IP; HSP-66Y; HSPC021; HSPC025; MSTP005; PAF67
          View more View less
          UniProt ID: (Human) Q9Y262, (Mouse) Q8QZY1
          View more View less
          Entrez Gene ID: (Human) 51386, (Mouse) 223691, (Rat) 300069
          View more View less
          Function(s)
          RNA binding translation initiation factor activity protein binding poly(A) RNA binding translation factor translation initiation factor translational protein
          Process(es)
          formation of cytoplasmic translation initiation complex cytoplasmic translational initiation translation translational initiation viral translational termination-reinitiation formation of translation preinitiation complex regulation of translational initiation
          Argentina flag icon
          Argentina
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline