    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
          • Primary Antibodies ›
          • PERK Antibodies
          Proteintech
          Phospho-PERK/EIF2AK3 (Thr982) Recombinant Rabbit Monoclonal Antibody (4E16), CoraLite® Plus 647
          View all (28) PERK antibodies
          Tech Support
          Cite Phospho-PERK/EIF2AK3 (Thr982) Recombinant Rabbit Monoclonal Antibody (4E16), CoraLite® Plus 647
          • Antibody Testing Data (1)
          Phospho-PERK/EIF2AK3 (Thr982) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          Phospho-PERK/EIF2AK3 (Thr982) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Phospho-PERK/EIF2AK3 (Thr982) Antibody (CL647-82534) in Flow
          1X10^6 HEK-293 cells untreated (dashed lines) or treated with CalycµLin A which intracellularly stained with 0.25 µg Coralite Plus 647 Phospho-PERK/EIF2AK3 (Thr982) Recombinant Antibody (CL647-82534, Clone: 4E16) (red), or 0.25 µg CoraLite Plus 647 Rabbit IgG Isotype Control RecAb (CL647-98136, Clone: 240953C9) (blue). Cells were fixed with 4% PFA and permeabilized with 90% MeOH. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Phospho-PERK/EIF2AK3 (Thr982) Antibody in Flow Cytometry (Flow)
          Product Details
          CL647-82534
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          0.25 µg per 10^6 cells
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Host/Isotype
          Rabbit / IgG
          Expression System
          CHO cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          4E16
          Immunogen
          Peptide
          Conjugate
          CoraLite® Plus 647 CoraLite® Plus 647 CoraLite® Plus 647
          • Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation
          Excitation/Emission Max
          654/674 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Target Information
          EIF2AK3 (PERK) is one of 4 kinases that specifically phosphorylate Ser51 of translation initiation factor eIF2-alpha in response to various environmental stresses, leading to a decrease in protein sythesis. In the case of EIF2AK3, signaling is initiated by misfolded proteins in the ER. Defects in EIF2AK3 are the cause of Wolcott-Rallison syndrome (WRS), also known as multiple epiphyseal dysplasia with early-onset diabetes mellitus. WRS is a rare autosomal recessive disorder, characterized by permanent neonatal or early infancy insulin-dependent diabetes and, at a later age, epiphyseal dysplasia, osteoporosis, growth retardation and other multisystem manifestations, such as hepatic and renal dysfunctions, mental retardation and cardiovascular abnormalities.
          Bioinformatics
          Protein Aliases: 2.7.11.1; Eukaryotic translation initiation factor 2-alpha kinase 3; HsPEK; OTTHUMP00000207187; OTTHUMP00000207188; Pancreatic eIF2-alpha kinase; PRKR-like endoplasmic reticulum kinase; Protein tyrosine kinase EIF2AK3
          View more View less
          Gene Aliases: AI427929; EIF2AK3; PEK; PERK; WRS
          View more View less
          UniProt ID: (Human) Q9NZJ5, (Mouse) Q9Z2B5
          View more View less
          Entrez Gene ID: (Human) 9451, (Mouse) 13666
          View more View less
          Function(s)
          protein kinase activity protein serine/threonine kinase activity eukaryotic translation initiation factor 2alpha kinase activity protein binding ATP binding enzyme binding protein phosphatase binding identical protein binding protein homodimerization activity Hsp90 protein binding nucleotide binding kinase activity transferase activity transferase activity, transferring phosphorus-containing groups eukaryotic initiation factor eIF2 binding non-receptor serine/threonine protein kinase
          Process(es)
          skeletal system development ossification angiogenesis chondrocyte development protein phosphorylation activation of cysteine-type endopeptidase activity involved in apoptotic process ER overload response endoplasmic reticulum organization positive regulation of vascular endothelial growth factor production positive regulation of gene expression regulation of translational initiation by eIF2 alpha phosphorylation negative regulation of translation peptidyl-serine phosphorylation calcium-mediated signaling insulin secretion bone mineralization endoplasmic reticulum unfolded protein response endocrine pancreas development negative regulation of myelination negative regulation of translational initiation in response to stress cellular response to amino acid starvation response to endoplasmic reticulum stress eiF2alpha phosphorylation in response to endoplasmic reticulum stress PERK-mediated unfolded protein response cellular response to glucose starvation positive regulation of transcription from RNA polymerase I promoter protein autophosphorylation insulin-like growth factor receptor signaling pathway protein homooligomerization regulation of endoplasmic reticulum stress-induced eIF2 alpha phosphorylation cellular response to cold positive regulation of protein localization to nucleus regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway response to manganese-induced endoplasmic reticulum stress translation regulation of translation response to unfolded protein lactation positive regulation of signal transduction negative regulation of gene expression phosphorylation regulation of fatty acid metabolic process pancreas development negative regulation of translation in response to stress positive regulation of protein binding SREBP signaling pathway negative regulation of apoptotic process fat cell differentiation intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress negative regulation of translation in response to endoplasmic reticulum stress positive regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway positive regulation of glutathione biosynthetic process positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong
          Your items have has been added!
          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline