    • See all product categories
  • Applications
    • Cell Analysis
    • Protein Biology
    • Lab Informatics
    • 360° CDMO and CRO Solutions
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
          • Primary Antibodies ›
          • SECTM1 Antibodies
          Proteintech
          SECTM1 Monoclonal Antibody (1C11B4), CoraLite® Plus 750
          View all (9) SECTM1 antibodies
          Tech Support
          Cite SECTM1 Monoclonal Antibody (1C11B4), CoraLite® Plus 750
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          SECTM1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SECTM1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          SECTM1 Antibody (CL750-60281) in WB
          Various lysates were subjected to SDS PAGE followed by western blot with CL750-60354 (CD99 antibody) at dilution of 1:500 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          SECTM1 Antibody in Western Blot (WB)
          Product Details
          CL750-60281
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:1,000
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgM
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1C11B4
          Immunogen
          SECTM1 fusion protein Ag14645
          Conjugate
          CoraLite® Plus 750 CoraLite® Plus 750 CoraLite® Plus 750
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          755/780 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Sequence of this protein is as follows: QNEGWDSPIC TEGVVSVSWG ENTVMSCNIS NAFSHVNIKL RAHGQESAIF NEVAPGYFSR DGWQLQVQGG VAQLVIKGAR DSHAGLYMWH LVGHQRNNRQ VTLEVSGAEP QSAPDTGFWP VPAVVTAVFI LLVALVMFAW YRCRCSQQRR EKKFFLLEPQ MKFAALRAGA QQGLSRASAE LWTPDSEPTP RPLALVFKPS PLGALELLSP QPLFPYAADP
          Target Information
          This gene encodes a transmembrane and secreted protein with characteristics of a type 1a transmembrane protein. It is found in a perinuclear Golgi-like pattern and thought to be involved in hematopoietic and/or immune system processes.
          Bioinformatics
          Protein Aliases: Protein K-12; Secreted and transmembrane protein 1; type 1a transmembrane protein; unnamed protein product
          View more View less
          Gene Aliases: K12; SECTM; SECTM1
          View more View less
          UniProt ID: (Human) Q8WVN6
          View more View less
          Entrez Gene ID: (Human) 6398
          View more View less
          Function(s)
          cytokine activity protein binding receptor agonist activity
          Process(es)
          positive regulation of T cell cytokine production immune response mesoderm development interleukin-2-mediated signaling pathway positive regulation of I-kappaB kinase/NF-kappaB signaling
          • Trademarks
          • Fair Trade Fair Trade
          Korea flag icon
          Korea
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910
          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783
          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)
          ISMS Logo
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline