    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Maltose Binding Protein Antibodies
          Proteintech
          MBP tag Monoclonal Antibody (4C6H4), CoraLite® Plus 750
          View all (16) Maltose Binding Protein antibodies
          Questions & Answers
          Cite MBP tag Monoclonal Antibody (4C6H4), CoraLite® Plus 750
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          MBP tag Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          MBP tag Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          MBP tag Antibody (CL750-66003) in WB
          HSC-T6 cells were subjected to SDS PAGE followed by western blot with CL750-66559 (SMAD1 antibody) at dilution of 1:1,000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          MBP tag Antibody in Western Blot (WB)
          Product Details
          CL750-66003
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000-1:8,000
          -
          Product Specifications
          Species Reactivity
          Tag
          Host/Isotype
          Mouse / IgG2a
          Class
          Monoclonal
          Type
          Antibody
          Clone
          4C6H4
          Immunogen
          MBP tag fusion protein Ag0942
          Conjugate
          CoraLite® Plus 750 CoraLite® Plus 750 CoraLite® Plus 750
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          755/780 nm
          Form
          Liquid
          Concentration
          1000 µg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Target Information
          The approach of epitope tagging offers an easy and universal strategy for the identification and purification of proteins derived by recombinant DNA technology. The insertion of a Maltose Binding Protein (MBP) tag creates a stable fusion product that does not interfere with the bioactivity of the protein or with the biodistribution of the MBP-tagged product. Cleavage by factor Xa separates MBP from its partner protein.
          Bioinformatics
          Protein Aliases: MBP; MBP Tag
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline