    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
          • Primary Antibodies ›
          • PPAR gamma Antibodies
          Proteintech
          PPAR Gamma Recombinant Rabbit Monoclonal Antibody (230374A3), CoraLite® Plus 750
          View all (25) PPAR gamma antibodies
          Tech Support
          Cite PPAR Gamma Recombinant Rabbit Monoclonal Antibody (230374A3), CoraLite® Plus 750
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          PPAR Gamma Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PPAR Gamma Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          PPAR Gamma Antibody (CL750-81490-5) in WB
          Various lysates were subjected to SDS PAGE followed by western blot with CL750-81490-5 (PPAR Gamma antibody) at dilution of 1:1,000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          PPAR Gamma Antibody in Western Blot (WB)
          PPAR Gamma Antibody in Flow Cytometry (Flow)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          PPAR Gamma Recombinant Rabbit Monoclonal Antibody (230374A3), CoraLite® Plus 750
          Product Details
          CL750-81490-5
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:2,000
          -
          Flow Cytometry (Flow)
          0.80 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications
          Species Reactivity
          Human, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          230374A3
          Immunogen
          PPAR Gamma fusion protein Ag10005
          3D Epitope / Immunogen
          Conjugate
          CoraLite® Plus 750 CoraLite® Plus 750 CoraLite® Plus 750
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          755/780 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 0.5% BSA, 50% glycerol
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: NKCQYCRFQK CLAVGMSHNA IRFGRMPQAE KEKLLAEISS DIDQLNPESA DLRALAKHLY DSYIKSFPLT KAKARAILTG KTTDKSPFVI YDMNSLMMGE DKIKFKHITP LQEQSKEVAI RIFQGCQFRS VEAVQEITEY AKSIPGFVNL DLNDQVTLLK YGVHEIIYTM LASLMNKDGV LISEGQGFMT REFLKSLRKP FGDFMEPKFE FAVKFNALEL DDSDLAIFIA VIILSGDRPG LLNVKPIEDI QDNLLQALEL QLKLNHPESS QLFAKLLQKM TDLRQIVTEH VQLLQVIKKT ETDMSLHPLL QEIYKDLY
          Target Information
          This gene encodes a member of the peroxisome proliferator-activated receptor (PPAR) subfamily of nuclear receptors. PPARs form heterodimers with retinoid X receptors (RXRs) and these heterodimers regulate transcription of various genes. Three subtypes of PPARs are known: PPAR-alpha, PPAR-delta, and PPAR-gamma. The protein encoded by this gene is PPAR-gamma and is a regulator of adipocyte differentiation. Additionally, PPAR-gamma has been implicated in the pathology of numerous diseases including obesity, diabetes, atherosclerosis and cancer. Alternatively spliced transcript variants that encode different isoforms have been described.
          Bioinformatics
          Protein Aliases: HUMPPARG; nuclear receptor; nuclear receptor subfamily 1 group C member 3; OTTHUMP00000185032; OTTHUMP00000185036; peroxisome proliferator; peroxisome proliferator activated receptor; peroxisome proliferator activator receptor, gamma; peroxisome proliferator-activated receptor-gamma 5; peroxisome proliferator-activated receptor-gamma splicing; PPAR gamma; PPAR-gamma; PPAR-gamma-1a; PPAR-gamma-1b; PPARy; unnamed protein product
          View more View less
          Gene Aliases: CIMT1; FPLD3; GLM1; NR1C3; PPARG1; PPARG2; PPARG5; PPARgamma; PPARgamma2
          View more View less
          UniProt ID: (Human) P37231, (Rat) O88275
          View more View less
          Entrez Gene ID: (Human) 5468, (Rat) 25664
          View more View less
          Function(s)
          transcription regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcription cofactor binding transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding nucleic acid binding DNA binding chromatin binding double-stranded DNA binding transcription factor activity, sequence-specific DNA binding RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding prostaglandin receptor activity protein binding zinc ion binding enzyme binding peptide binding identical protein binding sequence-specific DNA binding retinoid X receptor binding arachidonic acid binding DBD domain binding LBD domain binding WW domain binding alpha-actinin binding R-SMAD binding E-box binding STAT family protein binding DNA-binding transcription factor binding RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II core promoter sequence-specific DNA binding transcription coactivator binding protein phosphatase binding estrogen receptor binding promoter-specific chromatin binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter negative regulation of cytokine production placenta development response to dietary excess diet induced thermogenesis wound healing involved in inflammatory response negative regulation of acute inflammatory response regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter fatty acid metabolic process response to stress heart development regulation of blood pressure negative regulation of cell proliferation response to xenobiotic stimulus response to light stimulus response to mechanical stimulus hormone-mediated signaling pathway regulation of gene expression negative regulation of cardiac muscle hypertrophy positive regulation of gene expression negative regulation of gene expression negative regulation of macrophage derived foam cell differentiation positive regulation of cholesterol efflux negative regulation of cholesterol storage negative regulation of lipid storage negative regulation of smooth muscle cell migration long-chain fatty acid transport negative regulation of angiogenesis regulation of lipid metabolic process fatty acid oxidation cell differentiation monocyte differentiation negative regulation of cell growth BMP signaling pathway negative regulation of transforming growth factor beta receptor signaling pathway negative regulation of BMP signaling pathway intracellular receptor signaling pathway epithelial cell differentiation response to caffeine organ regeneration positive regulation of prostaglandin biosynthetic process response to nutrient levels response to food positive regulation of cholesterol transport positive regulation of intracellular cholesterol transport cellular response to insulin stimulus cellular response to hormone stimulus negative regulation of collagen biosynthetic process response to vitamin A response to lipid peroxisome proliferator activated receptor signaling pathway response to immobilization stress cellular response to platelet-derived growth factor stimulus glucose homeostasis response to starvation regulation of circadian rhythm mRNA transcription from RNA polymerase II promoter positive regulation of apoptotic process negative regulation of MAPK cascade negative regulation of blood vessel endothelial cell migration response to estrogen cell fate commitment fat cell differentiation regulation of fat cell differentiation positive regulation of fat cell differentiation negative regulation of osteoblast differentiation negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of fatty acid metabolic process positive regulation of transcription from RNA polymerase II promoter positive regulation of fatty acid oxidation regulation of hormone secretion retinoic acid receptor signaling pathway cell maturation negative regulation of smooth muscle cell proliferation positive regulation of oligodendrocyte differentiation positive regulation of protein secretion negative regulation of inflammatory response white fat cell differentiation brown fat cell differentiation positive regulation of phagocytosis, engulfment negative regulation of interferon-gamma-mediated signaling pathway positive regulation of SMAD protein import into nucleus negative regulation of SMAD protein import into nucleus positive regulation of adiponectin secretion response to linoleic acid cellular response to lithium ion cellular response to retinoic acid cellular response to vitamin E cellular response to prostaglandin stimulus cellular response to prostaglandin E stimulus cellular response to low-density lipoprotein particle stimulus cellular response to hyperoxia cellular response to hypoxia negative regulation of mitochondrial fission negative regulation of peptide hormone secretion cold-induced thermogenesis positive regulation of lipoprotein transport beige fat cell differentiation negative regulation of cellular response to insulin stimulus negative regulation of extracellular matrix assembly negative regulation of pri-miRNA transcription from RNA polymerase II promoter positive regulation of pri-miRNA transcription from RNA polymerase II promoter regulation of protein localization to plasma membrane negative regulation of cardiac muscle hypertrophy in response to stress negative regulation of cellular response to transforming growth factor beta stimulus positive regulation of adipose tissue development negative regulation of connective tissue replacement involved in inflammatory response wound healing negative regulation of vascular smooth muscle cell proliferation negative regulation of receptor signaling pathway via STAT positive regulation of vascular associated smooth muscle cell apoptotic process negative regulation of vascular endothelial cell proliferation negative regulation of pancreatic stellate cell proliferation positive regulation of apoptotic signaling pathway G-protein coupled receptor signaling pathway response to nutrient innate immune response positive regulation of lipid metabolic process regulation of transport negative regulation of multicellular organismal process lipid homeostasis nuclear receptor-mediated signaling pathway regulation of cellular response to insulin stimulus
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          India flag icon
          India
          Thermo Fisher Scientific India Pvt Ltd.
          Registered office address:
          402, 403, 404, Delphi 'B' Wing,
          Hiranandani Business Park,
          Powai, Mumbai – 400 076
          Email address: customer.reach@thermofisher.com
          Phone: 1800-2097001
          CIN: U73100MH2000PTC126872
          Invitrogen BioServices (India) Pvt Ltd
          Registered office address:
          First Technology Place, No 3, EPIP,
          Whitefield, Bangalore – 560 079
          Email address: customer.reach@thermofisher.com
          Phone: 1800-2097001
          CIN: U73100KA2004PTC035330
          Your items have has been added!
          Host server : magellan-search-green-5db85fbf84-frpqk:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch