    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • USP14 Antibodies
          Proteintech
          USP14 Recombinant Rabbit Monoclonal Antibody (3G12), CoraLite® Plus 750
          View all (25) USP14 antibodies
          Tech Support
          Cite USP14 Recombinant Rabbit Monoclonal Antibody (3G12), CoraLite® Plus 750
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          USP14 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          USP14 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          USP14 Antibody (CL750-82128) in WB
          Various lysates were subjected to SDS PAGE followed by western blot with CL750-82128 (USP14 antibody) at dilution of 1:8,000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          USP14 Antibody in Western Blot (WB)
          USP14 Antibody in Western Blot (WB)
          USP14 Antibody in Flow Cytometry (Flow)
          Product Details
          CL750-82128
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000-1:16,000
          -
          Flow Cytometry (Flow)
          0.40 µg/10^6 cells in a 100 µL suspension
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          CHO cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          3G12
          Immunogen
          USP14 fusion protein Ag5979
          Conjugate
          CoraLite® Plus 750 CoraLite® Plus 750 CoraLite® Plus 750
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          755/780 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Product Specific Information
          Immunogen sequence: MPLYSVTVK WGKEKFEGVE LNTDEPPMVF KAQLFALTGV QPARQKVMVK GGTLKDDDWG NIKIKNGMTL LMMGSADALP EEPSAKTVFV EDMTEEQLAS AMELPCGLTN LGNTCYMNAT VQCIRSVPEL KDALKRYAGA LRASGEMASA QYITAALRDL FDSMDKTSSS IPPIILLQFL HMAFPQFAEK GEQGQYLQQD ANECWIQMMR VLQQKLEAIE DDSVKETDSS SASAATPSKK KSLIDQFFGV EFETTMKCTE SEEEEVTKGK ENQLQLSCFI NQEVKYLFTG LKLRL
          Target Information
          This gene encodes a protein that is one of many involved in mechanisms to ensure proper chromosome segregation during cell division.
          Bioinformatics
          Protein Aliases: ataxia; Deubiquitinating enzyme 14; dUB-type TGT for deubiquitinating enzyme; tRNA-guanine transglycosylase, 60-kD subunit; Ubiquitin carboxyl-terminal hydrolase 14; ubiquitin specific peptidase 14 (tRNA-guanine transglycosylase); ubiquitin specific protease 14; ubiquitin specific protease 14 (tRNA-guanine transglycosylase); Ubiquitin thioesterase 14; ubiquitin thiolesterase 14; ubiquitin-specific processing protease 14; Ubiquitin-specific-processing protease 14; unnamed protein product
          View more View less
          Gene Aliases: 2610005K12Rik; 2610037B11Rik; AW107924; ax; C78769; nmf375; TGT; Ubp6; USP14
          View more View less
          UniProt ID: (Human) P54578, (Mouse) Q9JMA1
          View more View less
          Entrez Gene ID: (Human) 9097, (Mouse) 59025, (Rat) 291796
          View more View less
          Function(s)
          cysteine-type endopeptidase activity thiol-dependent ubiquitin-specific protease activity endopeptidase inhibitor activity protein binding peptidase activity cysteine-type peptidase activity hydrolase activity Lys63-specific deubiquitinase activity proteasome binding ubiquitinyl hydrolase activity thiol-dependent ubiquitinyl hydrolase activity cysteine protease protease protein modifying enzyme
          Process(es)
          immune system process proteolysis inflammatory response chemical synaptic transmission protein deubiquitination proteasome-mediated ubiquitin-dependent protein catabolic process innate immune response regulation of chemotaxis regulation of proteasomal protein catabolic process protein K48-linked deubiquitination negative regulation of ERAD pathway negative regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process ubiquitin-dependent protein catabolic process negative regulation of endopeptidase activity negative regulation of ER-associated ubiquitin-dependent protein catabolic process
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline