Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • CLAC-P Antibodies

          Invitrogen

          CLAC-P Polyclonal Antibody

          View all (8) CLAC-P antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CLAC-P Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          CLAC-P Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.
          CLAC-P Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CLAC-P Antibody (PA1-4308) in IHC

          Immunostaining of floating sections of neocortex of Alzheimer's disease brains (fixed with formalin for 24 hours) using Product # PA1-4308. At a high magnification, the CLAC-P (NC3 region) antibody stains senile plaques. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CLAC-P Antibody in Immunohistochemistry (IHC)
          CLAC-P Antibody in Western Blot (WB)
          CLAC-P Polyclonal Antibody

          Product Details

          PA1-4308

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:2,000
          -

          Immunohistochemistry (IHC)

          1:100-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide: D(430) Y N G N L H E A L Q R I T(443) of the NC3 region of human and murine CLAC-P
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS

          Contains

          0.02% sodium azide

          Storage conditions

          Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2276411

          Product Specific Information

          PA1-4308 detects CLAC-P (NC3 region) in human and mouse samples.

          PA1-4308 has been successfully used in Western blotting and immunohistochemistry procedures. By Western blot, this antibody detects a ~70 kDa protein representing CLAC-P (NC3 region) in stable HEK293T cell lines expressing CLAC-P (NC3 region). PA1-4308 does not cross react with COLXIII or COLXXIII proteins.

          PA1-4308 immunizing peptide corresponds to amino acid residues 430-443 of the NC3 region of human and mouse CLAC-P.

          Maintain at 4°C for 3 months or at -20°C for longer periods. Avoid freeze/thaw cycles.

          Target Information

          Alzheimer's disease (AD) is characterized by deposition of beta-amyloid as senile plaques and cerebral amyloid angiopathy. The principal component of AD amyloid is the amyloid beta peptide (A beta). Although production and deposition of A beta appear closely related to the pathogenesis of AD, current evidence suggest that A beta alone is not sufficient to cause neuronal death and symptoms of dementia. A component of senile plaque (SP), CLAC (collagen-like Alzheimer amyloid plaque component ) and its precursor CLAC-P are deposited with extracellular beta-amyloid and have been recently implicated in cell toxicity of beta-amyloid in AD brains. CLAC-P is a unique, membrane-bound collagen-like structure containing three Gly-X-Y repeat motifs and may define a novel class of neuronal collagens.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Alzheimer disease amyloid-associated protein; AMY; CLAC-P; CLAC-P type I; CLAC-P type II; col25a1; Collagen alpha-1(XXV) chain; collagen-like Alzheimer amyloid plaque component; collagenous Alzheimer amyloid plaque component; procollagen, type XXV, alpha 1

          View more View less

          Gene Aliases: 2700062B08Rik; AMY; CFEOM5; CLAC; CLAC-P; CLACP; COL25A1

          View more View less

          UniProt ID: (Human) Q9BXS0, (Mouse) Q99MQ5

          View more View less

          Entrez Gene ID: (Human) 84570, (Mouse) 77018

          View more View less

          Function(s)
          beta-amyloid binding heparin binding extracellular matrix structural constituent conferring tensile strength identical protein binding extracellular matrix structural protein
          Process(es)
          axonogenesis involved in innervation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline