Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • CLN3 Antibodies

          Abnova

          CLN3 Polyclonal Antibody, MaxPab™

          View all (8) CLN3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CLN3 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          CLN3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          CLN3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CLN3 Antibody (H00001201-D01P) in WB

          Western Blot analysis of CLN3 expression in transfected 293T cell line (H00001201-T02) by CLN3 MaxPab polyclonal antibody.

          Lane 1: CLN3 transfected lysate(47.60 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CLN3 Antibody in Western Blot (WB)
          CLN3 Antibody in Western Blot (WB)
          CLN3 Polyclonal Antibody, MaxPab™

          Product Details

          H00001201-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          CLN3 (NP_000077.1, 1 a.a. approximately 438 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MGGCAGSRRR FSDSEGEETV PEPRLPLLDH QGAHWKNAVG FWLLGLCNNF SYVVMLSAAH DILSHKRTSG NQSHVDPGPT PIPHNSSSRF DCNSVSTAAV LLADILPTLV IKLLAPLGLH LLPYSPRVLV SGICAAGSFV LVAFSHSVGT SLCGVVFASI SSGLGEVTFL SLTAFYPRAV ISWWSSGTGG AGLLGALSYL GLTQAGLSPQ QTLLSMLGIP ALLLASYFLL LTSPEAQDPG GEEEAESAAR QPLIRTEAPE SKPGSSSSLS LRERWTVFKG LLWYIVPLVV VYFAEYFINQ GLFELLFFWN TSLSHAQQYR WYQMLYQAGV FASRSSLRCC RIRFTWALAL LQCLNLVFLL ADVWFGFLPS IYLVFLIILY EGLLGGAAYV NTFHNIALET SDEHREFAMA ATCISDTLGI SLSGLLALPL HDFLCQLS

          Target Information

          CLN3 is a highly glycosylated, hydrophobic, 438-amino acid protein with 6 transmembrane domains. The CLN3 protein localizes to the lysosomal membrane and plays a role in lysosomal function. It may act as a chaperone involved in the folding and unfolding of other proteins, namely subunit C of the ATP synthase complex. Mutations in the CLN3 gene cause Batten disease, a recessively inherited neurodegenerative disorder of childhood caused by lysosomal accumulation of hydrophobic material, mainly ATP synthase subunit C. Batten disease is the most common form of a group of disorders known as neuronal ceroid lipofuscinoses (NCLs). Symptoms of Batten disease include progressive loss of vision, seizures, and psychomotor disturbances.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Batten disease; Batten disease protein; Battenin; ceroid-lipofuscinosis, neuronal 3; CLN 3; CLN3, battenin; MGC102840; Protein CLN3; unnamed protein product

          View more View less

          Gene Aliases: AI323623; BTN1; BTS; CLN3; JNCL; SLC29B1

          View more View less

          UniProt ID: (Human) Q13286, (Mouse) Q61124

          View more View less

          Entrez Gene ID: (Human) 1201, (Mouse) 12752

          View more View less

          Function(s)
          protein binding glycolipid transporter activity calcium-dependent protein binding glycolipid binding sulfatide binding membrane trafficking regulatory protein
          Process(es)
          action potential amino acid transport endocytosis receptor-mediated endocytosis lysosome organization lysosomal lumen acidification learning or memory associative learning cellular water homeostasis regulation of autophagy regulation of fibroblast migration macroautophagy regulation of autophagosome size protein processing cellular cation homeostasis actin cytoskeleton organization regulation of proteolysis regulation of synaptic transmission, GABAergic ionotropic glutamate receptor signaling pathway ceramide transport lysosomal lumen pH elevation renal potassium excretion amyloid precursor protein catabolic process positive regulation of Golgi to plasma membrane protein transport negative regulation of apoptotic process negative regulation of neuron apoptotic process blood vessel endothelial cell migration plasma membrane raft organization negative regulation of proteolysis glycerophospholipid biosynthetic process glycolipid transport vesicle transport along microtubule regulation of short-term neuronal synaptic plasticity positive regulation of pinocytosis neuromuscular process controlling balance regulation of cytosolic calcium ion concentration regulation of filopodium assembly regulation of cytoskeleton organization establishment of localization in cell regulation of synaptic transmission, glutamatergic membrane organization autophagosome-lysosome fusion regulation of biological quality regulation of protein processing protein localization to membrane protein localization to plasma membrane Golgi to lysosome transport phagosome-lysosome docking phagosome-lysosome fusion autophagosome maturation regulation of cellular response to osmotic stress regulation of arginine biosynthetic process regulation of autophagosome maturation regulation of protein localization to plasma membrane arginine transmembrane transport lysosomal protein catabolic process regulation of phagosome maturation regulation of modification of synaptic structure positive regulation of caveolin-mediated endocytosis globoside metabolic process positive regulation of protein phosphorylation cellular amino acid metabolic process ceramide metabolic process glucosylceramide metabolic process galactosylceramide metabolic process sphingomyelin metabolic process transport vacuolar transport regulation of gene expression arginine transport negative regulation of macroautophagy cell migration neurotransmitter metabolic process negative regulation of catalytic activity
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline