Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CNOT1 Antibodies

          Proteintech

          CNOT1 Polyclonal Antibody

          69 References
          View all (12) CNOT1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CNOT1 Polyclonal Antibody

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (17)
          CNOT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          CNOT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 17

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CNOT1 Antibody (14276-1-AP) in IHC (P)

          Immunohistochemistry of paraffin-embedded human heart tissue slide using 14276-1-AP ( CNOT1 Antibody) at dilution of 1:200 (under 10x lens). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CNOT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CNOT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CNOT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CNOT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CNOT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CNOT1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CNOT1 Antibody in Western Blot (WB)
          CNOT1 Antibody in Western Blot (WB)
          CNOT1 Antibody in Western Blot (WB)
          CNOT1 Antibody in Western Blot (WB)
          CNOT1 Antibody in Western Blot (WB)
          CNOT1 Antibody in Western Blot (WB)
          CNOT1 Antibody in Western Blot (WB)
          CNOT1 Antibody in Western Blot (WB)
          CNOT1 Antibody in Western Blot (WB)
          CNOT1 Antibody in Western Blot (WB)
          CNOT1 Antibody in Immunoprecipitation (IP)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          CNOT1 Polyclonal Antibody

          Product Details

          14276-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 55 publications 55 publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          -
          View 3 publications 3 publications

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 6 publications 6 publications

          ChIP assay (ChIP)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          CNOT1 Fusion Protein Ag5623 (1-354 aa encoded by BC040523)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.27 mg/mL

          Amount

          40.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          This is a rabbit polyclonal antibody raised against a region from the C-terminal CNOT1 of human origin. CNOT1 exists as many isoforms and molecular weight of isoforms are 267, 241, and 173 kDa.

          Immunogen sequence: MNLDSLSLA LSQISYLVDN LTKKNYRASQ QEIQHIVNRH GPEADRHLLR CLFSHVDFSG DGKSSGKDFH QTQFLIQECA LLITKPNFIS TLSYAIDNPL HYQKSLKPAP HLFAQLSKVL KLSKVQEVIF GLALLNSSSS DLRGFAAQFI KQKLPDLLRS YIDADVSGNQ EGGFQDIAIE VLHLLLSHLL FGQKGAFGVG QEQIDAFLKT LRRDFPQERC PVVLAPLLYP EKRDILMDRI LPDSGGVAKT MMESSLADFM QEVGYGFCAS IEECRNIIVQ FGVREVTAAQ VARVLGMMAR THSGLTDGIP LQSISAPGSG IWSDGKDKSD GAQAHTWNVE VLIDVLKELN PSLNF (1-354 aa encoded by BC040523)

          Target Information

          Scaffolding component of the CCR4-NOT complex which is one of the major cellular mRNA deadenylases and is linked to various cellular processes including bulk mRNA degradation, miRNA-mediated repression, translational repression during translational initiation and general transcription regulation. Additional complex functions may be a consequence of its influence on mRNA expression. Its scaffolding function implies its interaction with the catalytic complex module and diverse RNA-binding proteins mediating the complex recruitment to selected mRNA 3'UTRs. Involved in degradation of AU-rich element (ARE)-containing mRNAs probably via association with ZFP36. Mediates the recruitment of the CCR4-NOT complex to miRNA targets and to the RISC complex via association with TNRC6A, TNRC6B or TNRC6C. Acts as a transcriptional repressor. Represses the ligand-dependent transcriptional activation by nuclear receptors. Involved in the maintenance of embryonic stem (ES) cell identity. [UniProt]

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: adrenal gland protein AD-005; CCR4-associated factor 1; negative regulator of transcription subunit 1 homolog; NOT1 (negative regulator of transcription 1, yeast) homolog; unnamed protein product

          View more View less

          Gene Aliases: 6030411K04Rik; AD-005; CDC39; D830048B13; HPE12; NOT1; NOT1H; RGD1308009; VIBOS

          View more View less

          UniProt ID: (Human) A5YKK6, (Mouse) Q6ZQ08

          View more View less

          Entrez Gene ID: (Human) 23019, (Rat) 291841, (Mouse) 234594

          View more View less

          Function(s)
          RNA binding poly(A)-specific ribonuclease activity protein binding protein domain specific binding estrogen receptor binding retinoic acid receptor binding binding, bridging armadillo repeat domain binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay trophectodermal cell differentiation biological_process positive regulation of cytoplasmic mRNA processing body assembly negative regulation of translation negative regulation of intracellular estrogen receptor signaling pathway gene silencing by miRNA negative regulation of retinoic acid receptor signaling pathway positive regulation of nuclear-transcribed mRNA poly(A) tail shortening positive regulation of mRNA catabolic process positive regulation of nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay regulation of stem cell population maintenance nuclear-transcribed mRNA poly(A) tail shortening
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-65f6797749-ftzwf:80/100.66.76.64:80.
          git-commit: 74bf1af917bc7c7f736041a944cb80ce7b99a823
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.0-PolychromeNPI-91