Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: PKTAWRLDCA QLKEKNQWQL SADDLKKLKP GLEKDFLPLY FGWFLTKKSS ETLRKAGQVF LEELGNHKAF KKELRQFVPG DEPREKMDLV TYFGKRPPGV LHCTTKFCDY GKAPGAEEYA QQDVLKKSYS KAFTLTISAL FVTPKTTGAR VELSEQQLQL WPSDVDKLSP TDNLPRGSRA HITLGCAADV EAVQTGLDLL EILRQEKGGS RGEEVGELSR GKLYSLGNGR WMLTLAKNME VRAIFTGYYG KGKPVPTQGS RKGGALQSCT II
Positive Samples: HepG2, 293T, Rat brain, Mouse brain; Cellular Location: Lipid-anchor, Melanosome, Membrane
CNP may participate in RNA metabolism in the myelinating cell, CNP is the third most abundant protein in central nervous system myelin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2'',3''-cyclic nucleotide 3'' phosphodiesterase; 2', 3' cyclic nucleotide 3' phosphohydrolase; 2',3'-cyclic-nucleotide 3'-phosphodiesterase (EC 3.1.4.37); CNP; CNPase; cyclic nucleotide phosphodiesterase 1; unnamed protein product
Gene Aliases: CN37; Cnp-1; CNP1; CNPase; CNPF; CNPI; CNPII; HLD20
UniProt ID: (Human) P09543, (Rat) P13233, (Mouse) P16330
Entrez Gene ID: (Human) 1267, (Rat) 25275, (Mouse) 12799
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support