Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • Caspr2 Antibodies

          Abnova

          CNTNAP2 Polyclonal Antibody, MaxPab™

          View all (19) Caspr2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CNTNAP2 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          CNTNAP2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          CNTNAP2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CNTNAP2 Antibody (H00026047-B01P) in WB

          Western Blot analysis of CNTNAP2 expression in transfected 293T cell line by CNTNAP2 MaxPab polyclonal antibody. Lane 1: CNTNAP2 transfected lysate(146.41 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CNTNAP2 Antibody in Western Blot (WB)
          CNTNAP2 Polyclonal Antibody, MaxPab™

          Product Details

          H00026047-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          CNTNAP2 (NP_054860.1, 1 a.a. approximately 1331 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MQAAPRAGCG AALLLWIVSS CLCRAWTAPS TSQKCDEPLV SGLPHVAFSS SSSISGSYSP GYAKINKRGG AGGWSPSDSD HYQWLQVDFG NRKQISAIAT QGRYSSSDWV TQYRMLYSDT GRNWKPYHQD GNIWAFPGNI NSDGVVRHEL QHPIIARYVR IVPLDWNGEG RIGLRIEVYG CSYWADVINF DGHVVLPYRF RNKKMKTLKD VIALNFKTSE SEGVILHGEG QQGDYITLEL KKAKLVLSLN LGSNQLGPIY GHTSVMTGSL LDDHHWHSVV IERQGRSINL TLDRSMQHFR TNGEFDYLDL DYEITFGGIP FSGKPSSSSR KNFKGCMESI NYNGVNITDL ARRKKLEPSN VGNLSFSCVE PYTVPVFFNA TSYLEVPGRL NQDLFSVSFQ FRTWNPNGLL VFSHFADNLG NVEIDLTESK VGVHINITQT KMSQIDISSG SGLNDGQWHE VRFLAKENFA ILTIDGDEAS AVRTNSPLQV KTGEKYFFGG FLNQMNNSSH SVLQPSFQGC MQLIQVDDQL VNLYEVAQRK PGSFANVSID MCAIIDRCVP NHCEHGGKCS QTWDSFKCTC DETGYSGATC HNSIYEPSCE AYKHLGQTSN YYWIDPDGSG PLGPLKVYCN MTEDKVWTIV SHDLQMQTPV VGYNPEKYSV TQLVYSASMD QISAITDSAE YCEQYVSYFC KMSRLLNTPD GSPYTWWVGK ANEKHYYWGG SGPGIQKCAC GIERNCTDPK YYCNCDADYK QWRKDAGFLS YKDHLPVSQV VVGDTDRQGS EAKLSVGPLR CQGDRNYWNA ASFPNPSSYL HFSTFQGETS ADISFYFKTL TPWGVFLENM GKEDFIKLEL KSATEVSFSF DVGNGPVEIV VRSPTPLNDD QWHRVTAERN VKQASLQVDR LPQQIRKAPT EGHTRLELYS QLFVGGAGGQ QGFLGCIRSL RMNGVTLDLE ERAKVTSGFI SGCSGHCTSY GTNCENGGKC LERYHGYSCD CSNTAYDGTF CNKDVGAFFE EGMWLRYNFQ APATNARDSS SRVDNAPDQQ NSHPDLAQEE IRFSFSTTKA PCILLYISSF TTDFLAVLVK PTGSLQIRYN LGGTREPYNI DVDHRNMANG QPHSVNITRH EKTIFLKLDH YPSVSYHLPS SSDTLFNSPK SLFLGKVIET GKIDQEIHKY NTPGFTGCLS RVQFNQIAPL KAALRQTNAS AHVHIQGELV ESNCGASPLT LSPMSSATDP WHLDHLDSAS ADFPYNPGQG QAIRNGVNRN SAIIGGVIAV VIFTILCTLV FLIRYMFRHK GTYHTNEAKG AESAESADAA IMNNDPNFTE TIDESKKEWL I

          Target Information

          This gene encodes a member of the neurexin family which functions in the vertebrate nervous system as cell adhesion molecules and receptors. This protein, like other neurexin proteins, contains epidermal growth factor repeats and laminin G domains. In addition, it includes an F5/8 type C domain, discoidin/neuropilin- and fibrinogen-like domains, thrombospondin N-terminal-like domains and a putative PDZ binding site. This protein is localized at the juxtaparanodes of myelinated axons, and mediates interactions between neurons and glia during nervous system development and is also involved in localization of potassium channels within differentiating axons. This gene encompasses almost 1.5% of chromosome 7 and is one of the largest genes in the human genome. It is directly bound and regulated by forkhead box protein P2 (FOXP2), a transcription factor related to speech and language development. This gene has been implicated in multiple neurodevelopmental disorders, including Gilles de la Tourette syndrome, schizophrenia, epilepsy, autism, ADHD and mental retardation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Cell recognition molecule Caspr2; contactin associated protein like 2; Contactin-associated protein-like 2; DKFZp781D1846; homolog of Drosophila neurexin IV; unnamed protein product

          View more View less

          Gene Aliases: AUTS15; CASPR2; CDFE; CNTNAP2; KIAA0868; NRXN4; PTHSL1

          View more View less

          UniProt ID: (Human) Q9UHC6

          View more View less

          Entrez Gene ID: (Human) 26047

          View more View less

          Function(s)
          protease binding protein binding enzyme binding ion channel binding
          Process(es)
          startle response cell adhesion central nervous system development brain development learning neuron recognition cell proliferation transmission of nerve impulse striatum development limbic system development thalamus development cerebral cortex development adult behavior neuron projection development social behavior vocal learning clustering of voltage-gated potassium channels neuron projection morphogenesis prepulse inhibition superior temporal gyrus development protein localization to juxtaparanode region of axon vocalization behavior positive regulation of gap junction assembly
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline