Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MGPPGPPGPR GPSGAPGADG PQGPPGGIGN PGAVGEKGEP GEAGEPGLPG EGGPPGPKGE RGEKGESGPS GAAGPPGPKG PPGDDGPKGS PGPVGFPGDP GPPGEPGPAG QDGPPGDKGD DGEPGQTGSP GPTGEPGPSG PPGKRGPPGP AGPEGRQGEK GAKGEAGLEG PPGKTGPIGP QGAPGKPGPD GLRGIPGPVG EQGLPGSPGP DGPPGPMGPP GLPGLKGDSG PKGEKGHPGL IGLIGPPGEQ GEKGDRGLPG PQGSSGPKGE QGITGPSGPI GPPGPPGLPG PPGPKGAKGS SGPTGPKGEA GHPGPPGPPG PPGEVIQPLP IQASRTRRNI DASQLLDDGN GENYVDYADG MEEIFGSLNS LKLEIEQMKR PLGTQQNPAR TCKDLQLCHP DFPDGEYWVD PNQGCSRDSF KVYCNFTAGG STCVFPDKKS EGARITSWPK ENPGSWFSEF KRGKLLSYVD AEGNPVGVVQ MTFLRLLSAS AHQNVTYHCY QSVAWQDAAT GSYDKALRFL GSNDEEMSYD NNPYIRALVD GCATKKGYQK TVLEIDTPKV EQVPIVDIMF NDFGEASQKF GFEVGPACFM G
Type I collagen, the fibrillar collagen found in most connective tissues, and the only component of the collagen found in cartilage. Mutations in this gene are associated with osteogenesis imperfecta, Ehlers-Danlos syndrome, and idiopathic osteoporosis. Reciprocal translocations between chromosomes 17 and 22, where this gene and the gene for platelet-derived growth factor beta are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans, resulting from unregulated expression of the growth factor. Two transcripts, resulting from the use of alternate polyadenylation signals, have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Collagen alpha-1(V) chain; collagen, type V, alpha 1
Gene Aliases: COL5A1; EDSC
UniProt ID: (Human) P20908
Entrez Gene ID: (Human) 1289
Molecular Function: extracellular matrix structural protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support