Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MQALRVSQA LIRSFSSTAR NRFQNRVREK QKLFQEDNDI PLYLKGGIVD NILYRVTMTL CLGGTVYSLY SLGWASFPRN (1-79 aa encoded by BC002757 )
This protein is one of the nuclear-coded polypeptide chains of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: COX7A1; cytochrome c oxidase subunit VIIa 1; cytochrome c oxidase subunit VIIa polypeptide 1 (muscle); cytochrome c oxidase subunit VIIa-H; cytochrome c oxidase subunit VIIa-heart; cytochrome c oxidase subunit VIIa-M; cytochrome c oxidase subunit VIIa-muscle; unnamed protein product
Gene Aliases: COX7A; COX7AH; COX7AM
UniProt ID: (Human) P24310, (Mouse) P56392
Entrez Gene ID: (Human) 1346, (Mouse) 12865
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support