Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • Carboxypeptidase B1 Antibodies

          Proteintech

          CPB1 Polyclonal Antibody

          4 References
          View all (25) Carboxypeptidase B1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CPB1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (17)
          CPB1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          CPB1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 17

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CPB1 Antibody (12600-1-AP) in IHC (P)

          Immunohistochemical analysis of paraffin-embedded human pancreas tissue slide using (Product # 12600-1-AP) (CPB1 antibody) at dilution of 1:200 (under 10x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CPB1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CPB1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CPB1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CPB1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CPB1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CPB1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CPB1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CPB1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CPB1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CPB1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CPB1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CPB1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CPB1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CPB1 Antibody in Western Blot (WB)
          CPB1 Antibody in Western Blot (WB)
          CPB1 Antibody in Western Blot (WB)
          CPB1 Antibody in Western Blot (WB)
          CPB1 Polyclonal Antibody

          Product Details

          12600-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 2 publications 2 publications

          Immunohistochemistry (IHC)

          -
          View 2 publications 2 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          CPB1 Fusion Protein Ag3243 (73-417 aa encoded by BC015338)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.4 mg/mL

          Amount

          60 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: AEDTVTVEN VLKQNELQYK VLISNLRNVV EAQFDSRVRA TGHSYEKYNK WETIEAWTQQ VATENPALIS RSVIGTTFEG RAIYLLKVGK AGQNKPAIFM DCGFHAREWI SPAFCQWFVR EAVRTYGREI QVTELLDKLD FYVLPVLNID GYIYTWTKSR FWRKTRSTHT GSSCIGTDPN RNFDAGWCEI GASRNPCDET YCGPAAESEK ETKALADFIR NKLSSIKAYL TIHSYSQMMI YPYSYAYKLG ENNAELNALA KATVKELASL HGTKYTYGPG ATTIYPAAGG SDDWAYDQGI RYSFTFELRD TGRYGFLLPE SQIRATCEET FLAIKYVASY VLEHLY (73-417 aa encoded by BC015338)

          Target Information

          Three different procarboxypeptidases A and two different procarboxypeptidases B have been isolated. The B1 and B2 forms differ from each other mainly in isoelectric point. Carboxypeptidase B1 is a highly tissue-specific protein and is a useful serum marker for acute pancreatitis and dysfunction of pancreatic transplants. It is not elevated in pancreatic carcinoma.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Carboxypeptidase B; carboxypeptidase B1 (tissue); Pancreas-specific protein; pancreatic carboxypeptidase B; pancreatic carboxypeptidase B1; PASP; procarboxypeptidase B; protaminase; tissue carboxypeptidase B

          View more View less

          Gene Aliases: 0910001A18Rik; 1810063F02Rik; 2210008M23Rik; AI504870; CPB; CPB1; PASP; PCPB

          View more View less

          UniProt ID: (Human) P15086

          View more View less

          Entrez Gene ID: (Human) 1360, (Mouse) 76703, (Rat) 24271

          View more View less

          Function(s)
          carboxypeptidase activity metallocarboxypeptidase activity protein binding peptidase activity metallopeptidase activity zinc ion binding hydrolase activity metal ion binding metalloprotease
          Process(es)
          proteolysis biological_process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline