PA1-27799 detects CRIPT from human, mouse, and rat samples.
PA1-27799 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.
The PA1-27679 immunogen is a synthetic peptide corresponding to human HSPC139.
Involved in the cytoskeletal anchoring of DLG4 in excitatory synapses.