Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: GEGKRNQTT EMRPERQPQT REQDRAHQTG ETVTGSGTQT QAGATQTVEQ DSSHQTGRTS KQTQEATNDQ NRGTETHGQG RSQTSQAVTG GHAQIQAGTH TQTPTQTVEQ DSSHQTGSTS TQTQESTNGQ NRGTEIHGQG RSQTSQAVTG GHTQIQAGSH TETVEQDRSQ TVSHGGAREQ GQTQTQPGSG QRWMQVSNPE AGETVPGGQA QTGASTEPGR QEWSSTHPRR CVTEGQGDRQ PTVVGEEWVD DHSRETVILR LDQGNLHTSV SSAQGQDAAQ SEEKRGITAR ELYSYLRSTK P (196-495 aa encoded by BC030807)
This gene encodes a member of the 'fused gene' family of proteins, which contain N-terminus EF-hand domains and multiple tandem peptide repeats. The encoded protein contains two EF-hand Ca2+ binding domains in its N-terminus and two glutamine- and threonine-rich 60 amino acid repeats in its C-terminus. This gene, also known as squamous epithelial heat shock protein 53, may play a role in the mucosal/epithelial immune response and epidermal differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 53 kDa putative calcium-binding protein; 53 kDa squamous epithelial-induced stress protein; 58 kDa heat shock protein; squamous epithelial heat shock protein 53; tumor-related protein; unnamed protein product
Gene Aliases: C1orf10; Gm1015; PDRC1; SEP53
UniProt ID: (Human) Q9UBG3
Entrez Gene ID: (Human) 49860, (Mouse) 381457
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support