PA1-28122 detects CSRP2 from human, mouse, rat samples.
PA1-28122 has been successfully used in Western blot applications.
The PA1-28122 immunogen is: Recombinant fragment, corresponding to amino acids 50-125 of Human CSRP2.
CSRP2 is a member of the CSRP family of genes, encoding a group of LIM domain proteins, which may be involved in regulatory processes important for development and cellular differentiation. CRP2 contains two copies of the cysteine-rich amino acid sequence motif (LIM) with putative zinc-binding activity, and may be involved in regulating ordered cell growth. Other genes in the family include CSRP1 and CSRP3.