Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MRTPGPLPV LLLLLAGAPA ARPTPPTCYS RMRALSQEIT RDFNLLQVSE PSEPCVRYLP RLYLDIHNYC VLDKLRDFVA SPPCWKVAQV DSLKDKARKL YTIMNSFCRR DLVFLLDDCN ALEYPIPVTT VLPDRQR (1-136 aa encoded by BC031391)
C17 is a cytokine-like protein specifically expressed in bone marrow and cord blood mononuclear cells that bear the CD34 surface marker (Liu et al., 2000 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cytokine-like protein C17; NB4 apoptosis related protein
Gene Aliases: 4930443F05Rik; 4Cytl1; C17; C4orf4; Cyt1; Gm147; RGD1561846
UniProt ID: (Human) Q9NRR1
Entrez Gene ID: (Human) 54360, (Rat) 498392, (Mouse) 231162
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support