Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • Calmodulin 1/2/3 Antibodies

          Invitrogen

          Calmodulin 1/2/3 Polyclonal Antibody

          View all (5) Calmodulin 1/2/3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Calmodulin 1/2/3 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          Calmodulin 1/2/3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Calmodulin 1/2/3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Calmodulin 1/2/3 Antibody (PA5-104261) in IHC (P)

          Immunohistochemistry analysis of paraffin-embedded Calmodulin 3 in human brain tissues sections. Antigen retrieval was performed using citrate buffer. Samples were blocked with blocking buffer (1.5 hr, 22°C), incubated with Calmodulin 3 polyclonal antibody (Product # PA5-104261) using a dilution of 1:100 (1.5 hr, 22°C), followed by HRP conjugated goat anti-rabbit. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Calmodulin 1/2/3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Calmodulin 1/2/3 Antibody in Western Blot (WB)
          Calmodulin 1/2/3 Polyclonal Antibody

          Product Details

          PA5-104261

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          - -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human CALM1(Accession P0DP23), corresponding to amino acid residues V56-L106.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2853575

          Product Specific Information

          Antibody detects endogenous levels of total Calmodulin1/2/3.

          Target Information

          Calmodulin is a small, highly conserved calcium binding protein found in all eukaryotic cells. With the capacity to bind up to four calcium ions, this 17 kDa protein acts as an important intracellular receptor for regulatory calcium signals. As it binds calcium, calmodulin undergoes conformational changes which can increase its affinity for target proteins. It acts both directly, through interaction with key target enzymes, and indirectly, via specific kinases. Studies have found that calmodulin participates in the regulation of several biological processes including energy and biosynthetic metabolism, cell motility, exocytosis, cytoskeletal assembly, and intracellular modulation of both cAMP and calcium concentrations.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CALM2; calmodulin; Calmodulin 1 (phosphorylase kinase, delta); calmodulin 2 (phosphorylase kinase, delta); Calmodulin III; Calmodulin-1; Calmodulin-2; Calmodulin-3; caM; epididymis secretory protein Li 72; LP7057 protein; phosphorylase kinase delta; phosphorylase kinase subunit delta; phosphorylase kinase subunit delta 1; phosphorylase kinase subunit delta 2; phosphorylase kinase subunit delta 3; phosphorylase kinase, delta subunit; prepro-calmodulin 1; prepro-calmodulin 2; prepro-calmodulin 3; unnamed protein product

          View more View less

          Gene Aliases: 1500001E21Rik; AI256814; AI327027; AI461935; AL024000; AL024017; CALM; CALM1; CALM2; CALM3; CALML2; CAM; CAM1; CAM2; CAM3; CaMA; CAMB; CAMC; CAMI; CAMII; CAMIII; CPVT4; CPVT6; DD132; HEL-S-72; LQT14; LQT15; LQT16; PHKD; PHKD1; PHKD2; PHKD3; R75142; RNCAMIII

          View more View less

          UniProt ID: (Human) P0DP23, (Rat) P0DP29, (Mouse) P0DP26, (Mouse) P0DP27, (Rat) P0DP31

          View more View less

          Entrez Gene ID: (Human) 801, (Human) 805, (Human) 808, (Rat) 24242, (Mouse) 12313, (Rat) 50663, (Mouse) 12314, (Rat) 24244, (Mouse) 12315

          View more View less

          Function(s)
          calcium channel regulator activity calcium ion binding protein binding adenylate cyclase activator activity calcium channel inhibitor activity protein kinase binding titin binding protein serine/threonine kinase activator activity ion channel binding metal ion binding calcium-dependent protein binding protein phosphatase activator activity transporter inhibitor activity adenylate cyclase binding protein domain specific binding nitric-oxide synthase regulator activity type 3 metabotropic glutamate receptor binding phosphatidylinositol 3-kinase binding nitric-oxide synthase binding enzyme regulator activity thioesterase binding N-terminal myristoylation domain binding phospholipase binding protein N-terminus binding calmodulin-related
          Process(es)
          G2/M transition of mitotic cell cycle regulation of heart rate detection of calcium ion G-protein coupled receptor signaling pathway JAK-STAT cascade regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion autophagosome membrane docking substantia nigra development regulation of cytokinesis cellular response to interferon-beta positive regulation of JAK-STAT cascade regulation of calcium-mediated signaling response to calcium ion regulation of cardiac muscle contraction long-term synaptic potentiation regulation of ryanodine-sensitive calcium-release channel activity negative regulation of ryanodine-sensitive calcium-release channel activity cellular response to interferon-gamma calcineurin-mediated signaling regulation of cardiac muscle cell action potential organelle localization by membrane tethering presynaptic endocytosis negative regulation of high voltage-gated calcium channel activity regulation of cell communication by electrical coupling involved in cardiac conduction negative regulation of calcium ion export across plasma membrane mitochondrion-ER tethering response to amphetamine response to corticosterone establishment of protein localization to membrane establishment of protein localization to mitochondrial membrane regulation of synaptic vesicle endocytosis regulation of synaptic vesicle exocytosis activation of adenylate cyclase activity calcium-mediated signaling positive regulation of cyclic nucleotide metabolic process positive regulation of phosphoprotein phosphatase activity positive regulation of protein dephosphorylation positive regulation of DNA binding positive regulation of nitric-oxide synthase activity positive regulation of cyclic-nucleotide phosphodiesterase activity positive regulation of ryanodine-sensitive calcium-release channel activity regulation of store-operated calcium channel activity regulation of high voltage-gated calcium channel activity regulation of catalytic activity
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline