Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Calmodulin 2 Antibodies

          Invitrogen

          Calmodulin 2 Polyclonal Antibody

          1 Reference
          View all (3) Calmodulin 2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Calmodulin 2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          Calmodulin 2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Calmodulin 2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Calmodulin 2 Antibody (PA5-121166) in WB

          Western blot analysis of Calmodulin 2 in extracts of various cell lines. Samples were incubated with Calmodulin 2 Polyclonal antibody (Product # PA5-121166) using a dilution of 1:1,000, followed by HRP Goat Anti-Rabbit IgG (H+L) at a dilution of 1:10,000. Lysates/proteins: 25 µg per lane. Blocking buffer: 3% nonfat dry milk in TBST. Detection: ECL Basic Kit. Exposure time: 30s. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Calmodulin 2 Antibody in Western Blot (WB)
          Calmodulin 2 Polyclonal Antibody

          Product Details

          PA5-121166

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 1 publication 1 publication

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant fusion protein containing a sequence corresponding to amino acids 1-149 of human CALM2 (NP_001734.1).
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.08 mg/mL

          Amount

          108 µg

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2914738

          Product Specific Information

          Positive test controls include: NIH/3T3, MCF7, BT-474, NCI-H460, Mouse brain. The target is usually found in the following locations: Cytoplasm, cytoskeleton, spindle, spindle pole.

          Immunogen sequence: MADQLTEEQI AEFKEAFSLF DKDGDGTITT KELGTVMRSL GQNPTEAELQ DMINEVDADG NGTIDFPEFL TMMARKMKDT DSEEEIREAF RVFDKDGNGY ISAAELRHVM TNLGEKLTDE EVDEMIREAD IDGDGQVNYE EFVQMMTAK

          Target Information

          Calmodulin 2 (CALM2) is one of the three human genes encoding calmodulin, a highly conserved calcium-binding messenger protein critical for mediating various cellular processes. Calmodulin proteins are composed of two pairs of EF-hand motifs located at both the N- and C-termini, connected by a flexible central linker. Each EF-hand motif binds calcium ions, with the C-terminus possessing two high-affinity Ca2+ binding sites, while the N-terminus contains two low-affinity sites. Upon binding calcium, calmodulin undergoes a conformational change that enables it to interact with and regulate numerous target proteins, including kinases, phosphatases, ion channels, and other enzymes. Specifically, calcium/calmodulin (CaM) signaling pathways are involved in critical cellular functions such as muscle contraction, neurotransmitter release, and cellular growth. The interaction of CALM2 with different targets is essential for translating transient changes in intracellular calcium levels into specific cellular responses, making it a pivotal player in calcium signaling.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CALM2; calmodulin; calmodulin 2 (phosphorylase kinase, delta); Calmodulin-1; Calmodulin-2; Calmodulin-3; caM; LP7057 protein; phosphorylase kinase delta; phosphorylase kinase subunit delta; phosphorylase kinase subunit delta 2; prepro-calmodulin 2; unnamed protein product

          View more View less

          Gene Aliases: 1500001E21Rik; AL024017; CALM; CALM1; CALM2; CALM3; CALML2; CAM; CAM1; CAM2; CAM3; CAMB; CAMC; CAMII; CAMIII; LQT15; PHKD; PHKD2

          View more View less

          UniProt ID: (Human) P0DP23, (Mouse) P0DP27

          View more View less

          Entrez Gene ID: (Human) 805, (Mouse) 12314

          View more View less

          Function(s)
          calcium channel regulator activity calcium ion binding protein binding adenylate cyclase binding adenylate cyclase activator activity calcium channel inhibitor activity protein kinase binding titin binding protein serine/threonine kinase activator activity ion channel binding metal ion binding calcium-dependent protein binding protein phosphatase activator activity transporter inhibitor activity protein domain specific binding enzyme regulator activity nitric-oxide synthase regulator activity type 3 metabotropic glutamate receptor binding phosphatidylinositol 3-kinase binding protein N-terminus binding nitric-oxide synthase binding calmodulin-related
          Process(es)
          G2/M transition of mitotic cell cycle regulation of heart rate detection of calcium ion G-protein coupled receptor signaling pathway regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion substantia nigra development regulation of cytokinesis regulation of calcium-mediated signaling response to calcium ion regulation of cardiac muscle contraction long-term synaptic potentiation negative regulation of ryanodine-sensitive calcium-release channel activity calcineurin-mediated signaling presynaptic endocytosis regulation of cell communication by electrical coupling involved in cardiac conduction negative regulation of calcium ion export across plasma membrane regulation of catalytic activity
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline