Search
Search
Osenses
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: Cathelicidin antimicrobial peptide.
LL-37 is a cationic antimicrobial peptide derived from the precursor protein hCAP18, which is encoded by the CAMP gene. It is produced by epithelial cells and immune cells such as neutrophils and plays an important role in innate immunity. LL-37 exhibits broad antimicrobial activity against bacteria, viruses, and fungi, and also functions as an immunomodulatory molecule by influencing inflammation, chemotaxis, and wound healing. In addition to host defense, dysregulated LL-37 expression has been associated with inflammatory diseases and certain cancers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cathelicidin antimicrobial peptide; cathelin-related antimicrobial peptide
Gene Aliases: CRAMP
Entrez Gene ID: (Rat) 316010
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support