Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • Caveolin 3 Antibodies

          Proteintech

          Caveolin-3 Polyclonal Antibody

          1 Reference
          View all (16) Caveolin 3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Caveolin-3 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          Caveolin-3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Caveolin-3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Caveolin-3 Antibody (28358-1-AP) in IHC (P)

          Immunohistochemical analysis of paraffin-embedded mouse heart tissue slide using 28358-1-AP (Caveolin-3 antibody) at dilution of 1:200 (under 40x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Caveolin-3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Caveolin-3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Caveolin-3 Antibody in Western Blot (WB)

          Product Details

          28358-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:600
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Caveolin-3 Fusion Protein Ag28009

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          467 µg/mL

          Amount

          70.05 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice

          Product Specific Information

          Aliquoting is unnecessary for -20°C storage.

          Target Information

          Caveolin-3 (CAV3) acts as a scaffolding protein within the caveolar membrane. It interacts directly with G-protein alpha subunits and can functionally regulate their activity. Caveolin-3 may also regulate voltage-gated potassium channels. It also plays a role in the sarcolemma repair mechanism of both skeletal muscle and cardiomyocytes that permits rapid resealing of membranes disrupted by mechanical stress. Caveolin-3 also mediates the recruitment of CAVIN2 and CAVIN3 proteins to the caveolae. Mutations in the gene can result in HyperCKmia, rippling muscle disease 2, cardiomyopathy, long QT syndrome 9, sudden infant death syndrome, and myopathy.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CAV; Caveolin-3; cavolin 3; contains K15N mutation; M-caveolin; MGC126100; MGC126101; MGC126129; unnamed protein product

          View more View less

          Gene Aliases: AI385751; Cav-3; CAV3; LGMD1C; LQT9; M-cav; MPDT; RMD2; VIP-21; VIP21

          View more View less

          UniProt ID: (Human) P56539, (Rat) P51638, (Mouse) P51637

          View more View less

          Entrez Gene ID: (Human) 859, (Rat) 29161, (Mouse) 12391

          View more View less

          Function(s)
          calcium channel regulator activity protein binding sodium channel regulator activity potassium channel inhibitor activity enzyme binding alpha-tubulin binding ion channel binding macromolecular complex binding nitric-oxide synthase binding binding, bridging connexin binding protein C-terminus binding protein complex binding protein complex scaffold scaffold/adaptor protein
          Process(es)
          MAPK cascade response to hypoxia plasma membrane repair regulation of heart rate response to ischemia cardiac muscle hypertrophy triglyceride metabolic process calcium ion transport endocytosis plasma membrane organization actin filament organization positive regulation of cytosolic calcium ion concentration muscle organ development myoblast fusion regulation of heart contraction protein localization positive regulation of cell proliferation negative regulation of cardiac muscle hypertrophy positive regulation of myotube differentiation regulation of skeletal muscle contraction myotube differentiation regulation of transforming growth factor beta receptor signaling pathway cell differentiation positive regulation of microtubule polymerization cytoplasmic microtubule organization membrane raft organization T-tubule organization detection of muscle stretch regulation of signal transduction by receptor internalization regulation of membrane potential glucose homeostasis cholesterol homeostasis negative regulation of MAPK cascade negative regulation of cell size regulation of nerve growth factor receptor activity regulation of cytosolic calcium ion concentration nucleus localization establishment of localization in cell regulation of protein kinase B signaling regulation of calcium ion transport negative regulation of calcium ion transport cardiac muscle cell development regulation of cardiac muscle contraction negative regulation of sarcomere organization regulation of ventricular cardiac muscle cell membrane repolarization heart trabecula formation regulation of ventricular cardiac muscle cell membrane depolarization regulation of branching involved in mammary gland duct morphogenesis negative regulation of cell growth involved in cardiac muscle cell development caveola assembly protein localization to plasma membrane ventricular cardiac muscle cell action potential regulation of calcium ion import regulation of cardiac muscle cell action potential involved in regulation of contraction regulation of p38MAPK cascade regulation of membrane depolarization during cardiac muscle cell action potential negative regulation of membrane depolarization during cardiac muscle cell action potential negative regulation of potassium ion transmembrane transport regulation of sodium ion transmembrane transport cellular response to ionomycin negative regulation of protein localization to cell surface positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process positive regulation of caveolin-mediated endocytosis negative regulation of protein kinase activity cell growth negative regulation of MAP kinase activity muscle cell cellular homeostasis negative regulation of nitric-oxide synthase activity establishment of protein localization to plasma membrane negative regulation of potassium ion transmembrane transporter activity regulation of calcium ion transmembrane transporter activity regulation of sodium ion transmembrane transporter activity
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-kg5vd:80/100.66.78.64:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0