Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MAEPRGPVD HGVQIRFITE PVSGAEMGTL RRGGRRPAKD ARASTYGVAV RVQGIAGQPF VVLNSGEKGG DSFGVQIKGA NDQGASGALS SDLELPENPY SQVKGFPAPS QSSTSDEEPG AYWNGKLLRS HSQASLAGPG PVDPSNRSNS MLELAPKVAS PGSTIDTAPL SSVDSLINKF DSQLGGQARG RTGRRTRMLP PEQRKRSKSL DSRLPRDTFE ERERQSTNHW TSSTKYDNHV GTSKQPAQSQ NLSPLSGFSR SRQTQDWVLQ SFEEPRRSAQ DPTMLQFKST PDLLRDQQEA APPGSVDHMK ATIYGILREG SSESETSVRR KVSLVLEKMQ PLVMVSSGST K (1-350 aa encoded by BC146657)
Probably plays a role in the formation and regulation of the tight junction (TJ) paracellular permeability barrier. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: tight junction protein; unnamed protein product
Gene Aliases: 6330408J11Rik
UniProt ID: (Human) Q9P2M7
Entrez Gene ID: (Human) 57530, (Mouse) 70737, (Rat) 310655
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support