PA1-26579 detects Claspin from human samples.
PA1-26579 has been successfully used in Western blot procedures.
The PA1-26579 immunogen is a synthetic peptide corresponding to a portion of human human Claspin encoded within exon 15.
Xenopus claspin is an essential upstream regulator of checkpoint kinase 1 and triggers a checkpoint arrest of the cell cycle in the presence of DNA templates in Xenopus egg extracts. The human gene appears to be the homolog Xenopus claspin and its function has not been determined.