Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • Claudin 3 Antibodies

          Invitrogen

          Claudin 3 Polyclonal Antibody

          Advanced Verification
          82 Published Figures
          305 References
          View all (20) Claudin 3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Claudin 3 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          • Published Figures (82)
          • Advanced Verification (1)
          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 87

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Claudin 3 Antibody (34-1700) in ICC/IF

          Immunofluorescence analysis of Claudin-3 was performed using 70% confluent log phase NIH:OVCAR-3 cells. The cells were fixed with 4% paraformaldehyde for 5 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, and blocked with 2% BSA for 45 minutes at room temperature. The cells were labeled with Claudin 3 Polyclonal Antibody (Product # 34-1700) at 1:100 dilution in 0.1% BSA, incubated at 4 degree ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Claudin 3 Antibody in Immunohistochemistry (IHC)
          Claudin 3 Antibody in Western Blot (WB)
          Claudin 3 Antibody in Immunohistochemistry (IHC)
          Claudin 3 Antibody in Western Blot (WB)
          Claudin 3 Antibody in Western Blot (WB)
          Claudin 3 Antibody in Western Blot (WB)
          Claudin 3 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 3 Antibody in Western Blot (WB)
          Claudin 3 Antibody in Western Blot (WB)
          Claudin 3 Antibody in Western Blot (WB)
          Claudin 3 Antibody in Western Blot (WB)
          Claudin 3 Antibody in Western Blot (WB)
          Claudin 3 Antibody in Western Blot (WB)
          Claudin 3 Antibody in Western Blot (WB)
          Claudin 3 Antibody in Western Blot (WB)
          Claudin 3 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          Claudin 3 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 3 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 3 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 3 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Claudin 3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Claudin 3 Antibody in Immunohistochemistry, Immunohistochemistry (Paraffin) (IHC, IHC (P))
          Claudin 3 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Claudin 3 Antibody in Immunohistochemistry (IHC)
          Claudin 3 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Claudin 3 Antibody in Immunohistochemistry (IHC)
          Claudin 3 Antibody in Immunohistochemistry (IHC)
          Claudin 3 Antibody in Immunohistochemistry (IHC)
          Claudin 3 Antibody in Immunohistochemistry (IHC)
          Claudin 3 Antibody in Immunohistochemistry (IHC)
          Claudin 3 Antibody in Immunohistochemistry (IHC)
          Claudin 3 Antibody in Immunohistochemistry (IHC)
          Claudin 3 Antibody in Immunohistochemistry (IHC)
          Claudin 3 Antibody in Immunohistochemistry (IHC)
          Claudin 3 Antibody in Immunohistochemistry (IHC)
          Claudin 3 Antibody in Immunohistochemistry (IHC)
          Claudin 3 Antibody in Immunohistochemistry (IHC)
          Claudin 3 Antibody in Immunohistochemistry (IHC)
          Claudin 3 Antibody in Immunohistochemistry (IHC)
          Claudin 3 Antibody in Immunohistochemistry (IHC)
          Claudin 3 Antibody in Immunohistochemistry (IHC)
          Claudin 3 Antibody in Immunohistochemistry (IHC)
          Claudin 3 Antibody in Immunohistochemistry (IHC)
          Claudin 3 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Claudin 3 Antibody in Immunohistochemistry (IHC)
          Claudin 3 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Claudin 3 Antibody in Immunohistochemistry (IHC)
          Claudin 3 Antibody in Immunohistochemistry (IHC)
          Claudin 3 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Claudin 3 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Claudin 3 Antibody in Immunohistochemistry (IHC)
          Claudin 3 Antibody in Immunohistochemistry (IHC)
          Claudin 3 Antibody in Western Blot (WB)
          Claudin 3 Antibody in Western Blot (WB)
          Claudin 3 Antibody in Western Blot (WB)
          Claudin 3 Antibody
          Claudin 3 Polyclonal Antibody

          Product Details

          34-1700

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 122 publications 122 publications

          Immunohistochemistry (IHC)

          -
          View 74 publications 74 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          View 12 publications 12 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 10 publications 10 publications

          Immunocytochemistry (ICC/IF)

          1:100
          View 72 publications 72 publications

          ELISA (ELISA)

          Assay-dependent
          -

          Immunoprecipitation (IP)

          -
          View 2 publications 2 publications

          Proximity Ligation Assay (PLA) (PLA)

          -
          View 1 publication 1 publication

          Immunoelectrophoresis (IE)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 11 publications 11 publications
          Product Specifications

          Species Reactivity

          Dog, Human, Mouse

          Published species

          Amphibian, Bovine, Dog, Fish, Goat, Hamster, Human, Mouse, Pig, Plant, Rabbit, Rat, Rhesus monkey, Sheep, Virus, Xenopus, Zebrafish

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide derived from the C-terminal region of mouse Claudin-3
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.25 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533158

          Product Specific Information

          This antibody is specific for the 22 kDa Claudin-3 protein. Reactivity has been confirmed with mouse liver and lung homogenates, canine MDCK and human MCF-7 cell lysates by western blotting, and with formalin-fixed, paraffin-embedded (FFPE) human normal colon and colon cancer tissues by immunohistochemistry. For best results in immunohistochemistry with formalin-fixed, paraffin-embedded (FFPE) tissues, heat induced epitope retrieval (HIER) with citrate buffer, pH 6.0, is required prior to staining.

          Target Information

          Claudin 1-8 proteins are a family of transmembrane proteins associated with tight junctions. Tight junctions are specialized regions of cell to cell contact; made up of network of strands to act as a molecular gasket for preventing the leakage of ions, water etc. between cells. They are abundant in luminal epithelial sheets where they maintain epithelial cell polarity. Different tissues exhibit different Claudin composition. Differential expression of this gene has been observed in different types of malignancies, including breast cancer, ovarian cancer, hepatocellular carcinomas, urinary tumors, prostate cancer, lung cancer, head and neck cancers, thyroid carcinomas, etc. Alternatively spliced transcript variants encoding different isoforms have been found.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Claudin-3; Clostridium perfringens enterotoxin receptor 2; CPE-R 2; CPE-receptor 2; hRVP1; integral membrane protein claudin-3; Rat ventral prostate.1 protein homolog; unnamed protein product; ventral prostate.1 protein homolog; ventral prostate.1-like protein

          View more View less

          Gene Aliases: AI182374; C7orf1; CLDN3; CPE-R2; CPETR2; HRVP1; mRVP1; RVP1

          View more View less

          UniProt ID: (Dog) Q95KM5, (Human) O15551, (Mouse) Q9Z0G9

          View more View less

          Entrez Gene ID: (Dog) 403648, (Human) 1365, (Mouse) 12739

          View more View less

          Function(s)
          structural molecule activity identical protein binding transmembrane signaling receptor activity protein binding protein binding involved in cell-cell adhesion tight junction
          Process(es)
          response to hypoxia positive regulation of protein phosphorylation epithelial cell morphogenesis negative regulation of cell proliferation positive regulation of gene expression negative regulation of gene expression establishment of endothelial blood-brain barrier calcium-independent cell-cell adhesion via plasma membrane cell-adhesion molecules regulation of cell morphogenesis positive regulation of cell migration negative regulation of cell migration actin cytoskeleton reorganization cell junction maintenance negative regulation of wound healing positive regulation of wound healing regulation of membrane permeability positive regulation of bicellular tight junction assembly positive regulation of metallopeptidase activity retinal pigment epithelium development cell adhesion actin cytoskeleton organization cell junction assembly maintenance of blood-brain barrier cell-cell junction maintenance response to ethanol bicellular tight junction assembly response to Gram-positive bacterium regulation of transepithelial transport negative regulation of phosphate transmembrane transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline