FIGURE: 1 / 1
Clostridium perfringens is a spore forming, anaerobic bacteria widely distributed in the soil and the digestive tract of many domestic animals. Six Types (A, B, C, D, E and F) have been identified on the basis of the toxins produced, with Types B, C and D being the most commonly associated with disease in domestic animals.