Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • Collagen I Antibodies

          Invitrogen

          Collagen I Polyclonal Antibody

          Advanced Verification
          16 Published Figures
          45 References
          View all (12) Collagen I antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Collagen I Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          • Published Figures (16)
          • Advanced Verification (1)
          Collagen I Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Collagen I Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 20

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Collagen I Antibody (PA1-26204) in IHC (P)

          Immunohistochemistry of Collagen I was performed in formalin fixed, paraffin embedded normal kidney using Collagen I Polyclonal Antibody (Product # PA1-26204) at a dilution of 1:100 for 4 hours at room temperature. A peroxidase goat anti-rabbit was used as a secondary antibody at 1:10,000 for 45 min at RT. Localization: Distal tubules in normal kidney tissue. Note the absence of staining of glomeruli. Stain... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Collagen I Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Collagen I Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Collagen I Antibody in Western Blot (WB)
          Collagen I Antibody in Immunocytochemistry (ICC/IF)
          Collagen I Antibody in Western Blot (WB)
          Collagen I Antibody in Immunocytochemistry (ICC/IF)
          Collagen I Antibody in Immunocytochemistry (ICC/IF)
          Collagen I Antibody in Immunocytochemistry (ICC/IF)
          Collagen I Antibody in Immunohistochemistry (IHC)
          Collagen I Antibody in Immunohistochemistry (IHC)
          Collagen I Antibody in Western Blot (WB)
          Collagen I Antibody in Western Blot (WB)
          Collagen I Antibody in Western Blot (WB)
          Collagen I Antibody in Western Blot (WB)
          Collagen I Antibody in Western Blot (WB)
          Collagen I Antibody in Western Blot (WB)
          Collagen I Antibody in Western Blot (WB)
          Collagen I Antibody in Western Blot (WB)
          Collagen I Antibody in Flow Cytometry (Flow)
          Collagen I Antibody
          Collagen I Polyclonal Antibody

          Product Details

          PA1-26204

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:10,000
          View 16 publications 16 publications

          Immunohistochemistry (IHC)

          -
          View 14 publications 14 publications

          Multiplex Immunohistochemistry (mIHC)

          Assay-dependent
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          View 2 publications 2 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          View 10 publications 10 publications

          Flow Cytometry (Flow)

          Assay-dependent
          View 1 publication 1 publication

          ELISA (ELISA)

          1:5,000-1:50,000
          -

          Immunoprecipitation (IP)

          1:100
          -

          Dot blot (DB)

          Assay-dependent
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Bovine, Hamster, Human, Mouse, Pig, Rat

          Published species

          Dog, Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Collagen Type I from human and bovine placenta.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.15 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 8.0

          Contains

          0.01% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2260734

          Product Specific Information

          Some class-specific anti-collagens are specific for three-dimensional epitopes, resulting in diminished reactivity with denatured collagen or formalin-fixed, paraffin embedded tissues. This antibody reacts with most mammalian Type I collagens and has negligible cross-reactivity with Type II, III, IV, V or VI collagens. Cross-reactivity with other human serum proteins and non-collagen extracellular matrix proteins is neglible.|Western blot: this preparation results in a native conformation of the protein. It is recommend not to add 2-ME or DTT in lysis buffer and not boiling the sample.

          Target Information

          Collagen I is a Type I collagen with a triple helix structure comprised of two alpha-1 chains and one alpha-2 chain. Collagen I is a member of group I collagen (fibril-forming collagen) found in most connective tissues, and is abundant in bone, cornea, dermis and tendon. Mutations in the COL1A2 gene encoding the alpha-2 chain are associated with osteogenesis imperfecta types I-IV, Ehlers-Danlos syndrome type VIIB, recessive Ehlers-Danlos syndrome Classical type, idiopathic osteoporosis, and atypical Marfan syndrome. Symptoms associated with mutations in the COL1A2 gene which codes for the alpha-2 chain, however, tend to be less severe than mutations in the COL1A1 gene, reflecting the different role of alpha-2 chains in matrix integrity. Reciprocal translocations between chromosomes 17 and 22, where Collagen Type 1 genes and the gene for platelet derived growth factor beta are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans, resulting from unregulated expression of the growth factor. Two transcripts, resulting from the use of alternate polyadenylation signals, have been identified.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: alpha 1 (I) chain propeptide; alpha 2 type I procollagen; alpha 2(I) procollagen; alpha 2(I)-collagen; alpha-1 collagen type 1; alpha-1 collagen type I; alpha-1 type 1 collagen; Alpha-1 type I collagen; Alpha-2 type I collagen; alpha1 chain of type I collagen; alpha1(I) procollagen; alpha2 chain of type I collagen; COL1; COL1A1; COL1A1; matrix protein; collagen 1 alpha 2 chain; collagen 1 pro-alpha-2 chain; collagen alpha 1 chain type I; Collagen alpha-1(I) chain; collagen alpha-1(I) chain preproprotein; Collagen alpha-2(I) chain; collagen Col1-ColIII-1; collagen Col1-ColIII-2; collagen COL1A2; collagen I, alpha-2 polypeptide; collagen of skin, tendon and bone, alpha-1 chain; collagen of skin, tendon and bone, alpha-2 chain; collagen type I alpha 1; collagen, type 1, alpha 1; collagen, type I, alpha 1; collagen, type I, alpha 2; epididymis secretory sperm binding protein; osteogenesis imperfecta; pre-pro-alpha-2 type I collagen; pro-alpha-1 collagen type 1; procollagen type I, alpha 1; procollagen, type 1, alpha 1; procollagen, type I, alpha 1; procollagen, type I, alpha 2; Type I Collagen; type I proalpha 1; type I procollagen; type I procollagen alpha 1 chain; type I-alpha 1 collagen; unnamed protein product

          View more View less

          Gene Aliases: AA960264; AI325291; CAFYD; Col1a-1; Col1a-2; COL1A1; COL1A2; Cola-1; Cola-2; Cola1; Cola2; COLIA1; EDSARTH1; EDSARTH2; EDSC; EDSCV; I79_010913; Mov-13; Mov13; OI1; OI2; OI3; OI4; oim

          View more View less

          UniProt ID: (Human) P02452, (Bovine) Q3MHM2, (Human) P08123, (Bovine) P02465, (Rat) P02454, (Mouse) P11087, (Mouse) Q01149, (Rat) P02466

          View more View less

          Entrez Gene ID: (Pig) 100738123, (Human) 1277, (Bovine) 282187, (Human) 1278, (Bovine) 282188, (Pig) 100626716, (Rat) 29393, (Chinese hamster) 100759814, (Mouse) 12842, (Chinese hamster) 100766269, (Mouse) 12843, (Rat) 84352

          View more View less

          Function(s)
          protease binding extracellular matrix structural constituent protein binding extracellular matrix structural constituent conferring tensile strength identical protein binding metal ion binding platelet-derived growth factor binding protein binding, bridging SMAD binding molecular_function extracellular matrix structural protein
          Process(es)
          skeletal system development ossification blood vessel development osteoblast differentiation intramembranous ossification endochondral ossification response to nutrient visual perception sensory perception of sound response to xenobiotic stimulus response to mechanical stimulus positive regulation of epithelial to mesenchymal transition negative regulation of cell-substrate adhesion protein transport collagen fibril organization positive regulation of cell migration response to nutrient levels response to estradiol response to insulin collagen biosynthetic process protein localization to nucleus tooth mineralization collagen-activated tyrosine kinase receptor signaling pathway response to hydrogen peroxide response to peptide hormone skin development skin morphogenesis cellular response to fibroblast growth factor stimulus positive regulation of transcription, DNA-templated animal organ development response to steroid hormone skeletal system morphogenesis embryonic skeletal system development system development response to cAMP response to hyperoxia face morphogenesis bone trabecula formation cartilage development involved in endochondral bone morphogenesis cellular response to amino acid stimulus cellular response to mechanical stimulus cellular response to retinoic acid cellular response to vitamin E cellular response to glucose stimulus cellular response to tumor necrosis factor cellular response to epidermal growth factor stimulus cellular response to transforming growth factor beta stimulus positive regulation of canonical Wnt signaling pathway response to fluoride cellular response to fluoride transforming growth factor beta receptor signaling pathway Rho protein signal transduction regulation of blood pressure bone mineralization collagen metabolic process odontogenesis protein heterotrimerization extracellular matrix assembly response to corticosteroid wound healing response to drug tooth eruption cellular response to organic substance response to norepinephrine cellular response to thyroid hormone stimulus
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-p58g2:80/100.66.72.198:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0