Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Search Thermo Fisher Scientific
PA5-16222 targets Cyclin B1 in immunohistochemistry (paraffin), immunoprecipitation, and Western blot applications and shows reactivity with mouse, Rat, and Human samples.
The PA5-16222 immunogen is a synthetic peptide derived from the C-terminus of human cyclin B1.
Cyclins bind to and regulate the activity of the Cyclin dependent protein kinases (CDKs). Cyclin B1 or CCNB1 is a regulatory protein involved in mitosis. It is essential for the control of the cell cycle at the G2/M transition. Cyclin B1 complexes with p34 (cdc2) to form the maturation-promoting factor (MPF). Two alternative transcripts have been found, a constitutively expressed transcript and a cell cycle-regulated transcript, that is expressed predominantly during G2/M phase. Cyclin B1 is not ubiquitinated during G2/M phase, resulting in its steady accumulation during G2 phase, followed by abrupt APC dependent destruction at the end of mitosis. Destruction of Cyclin B1 is required for cell cycle progression. Cyclin B1 is overexpressed in various cancers, including breast, prostate, and non-small cell lung cancer.