Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • New Products
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • DAB2IP Antibodies

          Proteintech

          DAB2IP Polyclonal Antibody

          View all (9) DAB2IP antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite DAB2IP Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          DAB2IP Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          DAB2IP Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          DAB2IP Antibody (30020-1-AP) in WB

          Various lysates were subjected to SDS PAGE followed by western blot with 30037-1-AP (SCNN1G antibody) at dilution of 1:1,000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          DAB2IP Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          DAB2IP Polyclonal Antibody

          Product Details

          30020-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:8,000
          -

          ELISA (ELISA)

          Assay-dependent
          -

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          DAB2IP fusion protein Ag31340
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          450 µg/mL

          Amount

          67.5 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice

          Product Specific Information

          Sequence of this protein is as follows: RGLGDSGSEG HSSLSSHSNS EELAAAAKLG SFSTAAEELA RRPGELARRQ MSLTEKGGQP TVPRQNSAGP QRRIDQPPPP PPPPPPAPRG RTPPNLLSTL QYPRPSSGTL ASASPDWVGP STRLRQQSSS SKGDSPELKP RAVHKQGPSP VSPNALDRTA AWLLTMNAQL LEDEGLGPDP PHRDRLRSKD ELSQAEKDLA VLQDKLRIST KK

          Target Information

          DAB2IP is a Ras GTPase-activating protein (GAP) that acts as a tumor suppressor. The DAB2IP gene is inactivated by methylation in prostate and breast cancers.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ASK-interacting protein 1; ASK1-interacting protein 1; DAB2 interaction protein; DAB2-interacting protein; DIP1/2; disabled 2 interacting protein long form; DOC-2/DAB-2 interactive protein; DOC-2/DAB2 interactive protein; DOC2/DAB2 interactive protein; MLL fusion partner; nGAP-like protein; ras GTPase activating protein; Ras-GTPase activating protein; RasGAP; unnamed protein product

          View more View less

          Gene Aliases: 2310011D08Rik; AF9Q34; AIP-1; AIP1; DIP1/2; mKIAA1743

          View more View less

          UniProt ID: (Human) Q5VWQ8, (Mouse) Q3UHC7, (Rat) Q6P730

          View more View less

          Entrez Gene ID: (Human) 153090, (Mouse) 69601, (Rat) 192126

          View more View less

          Function(s)
          GTPase activator activity death receptor binding protein binding SH3 domain binding kinase binding protein kinase binding mitogen-activated protein kinase kinase binding mitogen-activated protein kinase kinase kinase binding phosphatidylinositol-3-phosphate binding signaling adaptor activity Toll-like receptor 4 binding phosphatidylinositol 3-kinase regulatory subunit binding identical protein binding protein homodimerization activity vascular endothelial growth factor receptor 2 binding protein serine/threonine kinase activator activity phosphatidylinositol 3-kinase binding macromolecular complex binding cadherin binding protein phosphatase 2A binding phosphatidylinositol-4-phosphate binding 14-3-3 protein binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter negative regulation of cell proliferation extrinsic apoptotic signaling pathway via death domain receptors negative regulation of endothelial cell migration negative regulation of epithelial cell migration negative regulation of epithelial to mesenchymal transition positive regulation of neuron projection development negative regulation of angiogenesis cell motility involved in cerebral cortex radial glia guided migration layer formation in cerebral cortex protein catabolic process negative regulation of cell growth negative regulation of vascular endothelial growth factor receptor signaling pathway positive regulation of protein complex assembly tumor necrosis factor-mediated signaling pathway negative regulation of toll-like receptor 4 signaling pathway negative regulation of GTPase activity tube formation intracellular signal transduction cellular response to vascular endothelial growth factor stimulus vascular endothelial growth factor receptor-2 signaling pathway negative regulation of epidermal growth factor receptor signaling pathway negative regulation of protein catabolic process positive regulation of apoptotic process regulation of GTPase activity regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of I-kappaB kinase/NF-kappaB signaling negative regulation of I-kappaB kinase/NF-kappaB signaling regulation of protein complex assembly negative regulation of MAPK cascade positive regulation of MAPK cascade positive regulation of GTPase activity positive regulation of protein catabolic process negative regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter positive regulation of JNK cascade negative regulation of Ras protein signal transduction negative regulation of fibroblast proliferation neuron projection morphogenesis positive regulation of epithelial cell proliferation negative regulation of epithelial cell proliferation regulation of cell cycle negative regulation of protein kinase B signaling intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress negative regulation of G0 to G1 transition negative regulation of ERK1 and ERK2 cascade cellular response to lipopolysaccharide cellular response to interleukin-1 cellular response to tumor necrosis factor cellular response to epidermal growth factor stimulus negative regulation of canonical Wnt signaling pathway positive regulation of synapse maturation lipid droplet formation positive regulation of dendrite development regulation of p38MAPK cascade negative regulation of vascular endothelial growth factor signaling pathway negative regulation of NIK/NF-kappaB signaling positive regulation of proteasomal protein catabolic process regulation of intracellular signal transduction positive regulation of neuron migration positive regulation of apoptotic signaling pathway cellular response to unfolded protein endothelial cell apoptotic process positive regulation of IRE1-mediated unfolded protein response
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch