PA1-27654 detects DAPP1 from human samples.
PA1-27654 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.
The PA1-27511 immunogen is a synthetic peptide corresponding to human Dual adapter for phosphotyrosine and 3-phosphotyrosine and 3-phosphoinositide.
May act as a B-cell-associated adapter that regulates B-cell antigen receptor (BCR)-signaling downstream of PI3K.