Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • DCP Antibodies

          Bioss

          DCP Polyclonal Antibody

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite DCP Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          DCP Polyclonal Antibody

          Product Details

          BS-1037R

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:100-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          Flow Cytometry (Flow)

          1:20-1:100
          -

          ELISA (ELISA)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KLH conjugated synthetic peptide derived from human DCP, amino acids 31-130.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          0.01M TBS, pH 7.4, with 50% glycerol, 1% BSA

          Contains

          0.02% ProClin 300

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          Protein Induced by Vitamin K Absence or Antagonist-II (PIVKA-II), also known as des-gamma-carboxy prothrombin (DCP), is an abnormal form of prothrombin produced primarily by liver tissues under conditions of vitamin K deficiency or antagonism. This immature prothrombin variant lacks sufficient coagulation activity and is prominently overexpressed in hepatocellular carcinoma (HCC) cells, serving as a crucial biomarker for screening and diagnosing HCC. PIVKA-II also appears in some cases of gastric cancer, albeit less frequently. Elevated serum levels of PIVKA-II are not exclusive to malignancies; they can also be seen in non-HCC conditions, which necessitates differential diagnostic strategies. The utility of PIVKA-II is further underscored by its ability to predict extrahepatic metastases in early HCC, supporting its role in comprehensive patient evaluation and monitoring.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: coagulation factor 2; Coagulation factor II; PIVKA-II; prepro-coagulation factor II; Protein Induced by Vitamin K Absence or Antagonist-II; Prothrombin; prothrombin B-chain; thrombin; thrombin factor II; thrombin precursor; unnamed protein product

          View more View less

          Gene Aliases: Cf-2; Cf2; F2; FII; PT; RPRGL2; THPH1

          View more View less

          UniProt ID: (Mouse) P19221, (Human) P00734

          View more View less

          Entrez Gene ID: (Mouse) 14061, (Rat) 29251, (Human) 2147

          View more View less

          Function(s)
          serine-type endopeptidase activity receptor binding calcium ion binding peptidase activity thrombospondin receptor activity lipopolysaccharide binding endopeptidase activity protein binding growth factor activity heparin binding serine-type peptidase activity hydrolase activity receptor agonist activity protease serine protease protein modifying enzyme
          Process(es)
          positive regulation of protein phosphorylation proteolysis acute-phase response cell surface receptor signaling pathway blood coagulation positive regulation of cell proliferation regulation of cell shape response to wounding regulation of gene expression positive regulation of phosphatidylinositol 3-kinase signaling response to inactivity platelet activation positive regulation of blood coagulation positive regulation of cell growth positive regulation of collagen biosynthetic process wound healing fibrinolysis negative regulation of proteolysis negative regulation of astrocyte differentiation positive regulation of release of sequestered calcium ion into cytosol regulation of cytosolic calcium ion concentration cellular response to mechanical stimulus positive regulation of phospholipase C-activating G-protein coupled receptor signaling pathway positive regulation of reactive oxygen species metabolic process G-protein coupled receptor signaling pathway hemostasis negative regulation of platelet activation regulation of blood coagulation negative regulation of blood coagulation positive regulation of insulin secretion positive regulation of JAK-STAT cascade regulation of body fluid levels cytolysis by host of symbiont cells positive regulation of protein kinase B signaling negative regulation of fibrinolysis antimicrobial humoral immune response mediated by antimicrobial peptide thrombin receptor signaling pathway neutrophil mediated killing of gram-negative bacterium negative regulation of cytokine production involved in inflammatory response positive regulation of protein localization to nucleus ligand-gated ion channel signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline