Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: IPQVTSDGK RLQVIVCSAT LHSFDVKKLS EKIMHFPTWV DLKGEDSVPD TVHHVVVPVN PKTDRLWERL GKSHIRTDDV HAKDNTRPGA NSPEMWSEAI KILKGEYAVR AIKEHKMDQA IIFCRTKIDC DNLEQYFIQQ GGGPDKKGHQ FSCVCLHGDR KPHERKQNLE RFKKGDVRFL ICTDVAARGI DIHGVPYVIN VTLPDEKQNY VHRIGRVGRA ERMGLAISLV ATEKEKVWYH VCSSRGKGCY NTRLKEDGGC TIWYNEMQLL SEIEEHLNCT ISQVEPDIKV PVDEFDGKVT YGQKRAAGGG SYKGHVDILA PTVQELAALE KEAQTSFLHL GYLPNQLFRT F (391-740 aa encoded by BC012132)
DDX1 (ATP-dependent RNA helicase DDX1) acts as an ATP-dependent RNA helicase. It is able to unwind both RNA-RNA and RNA-DNA duplexes. DDX1 possesses 5' single-stranded RNA overhand nuclease activity and ATPase activity on various RNA, but not DNA polynucleotides. DDX1, together with RELA, acts as a coactivator to enhance NF-kappa-b-mediated transcriptional activation. It also acts as a positive transcriptional regulator of cyclin CCND2 expression. It binds to the cyclin CCND2 promoter region. DDX1 is a component of a multi-helicase-TICAM1 complex that acts as a cytoplasmic sensor of viral double stranded RNA and plays a role in activation of a cascade of antiviral responses including the induction of proinflammatory cytokines via the adapter molecule TICAM1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Asp-Glu-Ala-Asp box; DEAD (Asp-Glu-Ala-Asp) box helicase 1; DEAD (Asp-Glu-Ala-Asp) box polypeptide 1; DEAD box polypeptide 1; DEAD box protein 1; DEAD box protein retinoblastoma; DEAD box-1; DEAD-box RNA helicase DDX1; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 1; DEAD/H-box helicase 1; unnamed protein product
Gene Aliases: DBP-RB; TSLIG6; UKVH5d
UniProt ID: (Human) Q92499, (Rat) Q641Y8, (Mouse) Q91VR5
Entrez Gene ID: (Human) 1653, (Rat) 84474, (Mouse) 104721
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support