Search
Search
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Sequence of this protein is as follows: MANETQKVGA IHFPFPFTPY SIQEDFMAEL YRVLEAGKIG IFESPTGTGK SLSLICGALS WLRDFEQKKR EEEARLLETG TGPLHDEKDE SLCLSSSCEG AAGTPRPAGE PAWVTQFVQK KEERDLVDRL KAEQARRKQR EERLQQLQHR VQLKYAAKRL RQEEEERENL LRLSREMLET GPEAERLEQL ESGEEELVLA EYESDEEKKV ASRVDEDEDD LEEEHITKIY YCSRTHSQLA QFVHEVKKSP FGKDVRLVSL GSRQNLCVNE DVKSLGSVQL INDRCVDMQR SRHEKKKGAE EEKPKRRRQE KQAACPFYNH EQMGLLRDEA LAEVKDMEQL LALGKEARAC PYYGSRLAIP AAQLVVLPYQ MLLHAATRQA AGIRLQDQVV IIDEAHNLID TITGMHSVEV SGSQLCQAHS QLLQYVERYG KRLKAKNLMY LKQILYLLEK FVAVLGGNIK QNPNTQSLSQ TGTELKTIND FLFQSQIDNI NLFKVQRYCE KSMISRKLFG FTERYGAVFS SREQPKLAGF QQFLQSLQPR TTEALAAPAD ESQASTLRPA SPLMHIQGFL AALTTANQDG RVILSRQGSL SQSTLKFLLL NPAVHFAQVV KECRAVVIAG GTMQPVSDFR QQLLACAGVE AERVVEFSCG HVIPPDNILP LVICSGISNQ PLEFTFQKRE LPQMMDEVGR ILCNLCGVVP GGVVCFFPSY EYLRQVHAHW EKGGLLGRLA ARKKIFQEPK SAHQVEQVLL AYSRCIQACG QERGQVTGAL LLSVVGGKMS EGINFSDNLG RCVVMVGMPF PNIRSAELQE KMAYLDQTLS PRPGTPREGS GGEPVHEGRQ PVHRQGHQAP EGFCQRSAPG PAICPAPCPG QAAGLDPSPC GGQSYLWPRH CCCAEVSPGE VGLFLMGNHA TAWRRALPLS CPLETVFVVG VVCGDPVTKV KPRRRVWSPE CCQDPGTGVS SRRRKWGNPE
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. DDX11 encodes a DEAD box protein, which is an enzyme that possesses both ATPase and DNA helicase activities. DDX11 is a homolog of the yeast CHL1 gene, and may function to maintain chromosome transmission fidelity and genomestability.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CHL1-like helicase homolog; CHL1-related helicase gene-1; CHL1-related protein 1; DEAD/H (Asp-Glu-Ala-Asp/His) box helicase 11; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 11 (CHL1-like helicase homolog, S. cerevisiae); DEAD/H box protein 11; DNA 5'-3' helicase DDX11; hCHLR1; keratinocyte growth factor-regulated gene 2 protein; KRG-2; probable ATP-dependent DNA helicase DDX11; probable ATP-dependent RNA helicase DDX11; unnamed protein product
Gene Aliases: CHL1; CHLR1; KRG2; WABS
UniProt ID: (Human) Q96FC9
Entrez Gene ID: (Human) 1663
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support