Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: TLTLVHQAP ARILHKKHTK KMDKTAKLDL LMQKIGMRGK PKVIDLTRNE ATVETLTETK IHCETDEKDF YLYYFLMQYP GRSLVFANSI SCIKRLSGLL KVLDIMPLTL HACMHQKQRL RNLEQFARLE DCVLLATDVA ARGLDIPKVQ HVIHYQVPRT SEIYVHRSGR TARATNEGLS LMLIGPEDVI NFKKIYKTLK KDEDIPLFPV QTKYMDVVKE RIRLARQIEK SEYRNFQACL HNSWIEQAAA ALEIELEEDM YKGGKADQQE ERRRQKQMKV LKKELRHLLS QPLFTESQKT KYPTQSGKPP LLVSAPSKSE SALSCLSKQK KKKTKKPKEP QPEQPQPSTS AN (509-859 aa encoded by BC008847)
METAP1 removes the amino-terminal methionine from nascent proteins. METAP1 is required for normal progression through the cell cycle.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADRH; ATP-dependent RNA helicase DDX24; DEAD (Asp-Glu-Ala-Asp) box helicase 24; DEAD (Asp-Glu-Ala-Asp) box polypeptide 24; DEAD box protein 24; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 13 (RNA helicase A); DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 24; S. cerevisiae CHL1-like helicase; unnamed protein product
Gene Aliases: 1700055J08Rik; 2510027P10Rik; AI649272; DDX24
UniProt ID: (Human) Q9GZR7, (Mouse) Q9ESV0
Entrez Gene ID: (Human) 57062, (Rat) 373065, (Mouse) 27225
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support