Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • DECR2 Antibodies

          Abnova

          DECR2 Polyclonal Antibody, MaxPab™

          View all (8) DECR2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite DECR2 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          DECR2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          DECR2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          DECR2 Antibody (H00026063-B01P) in WB

          Western Blot analysis of DECR2 expression in transfected 293T cell line (H00026063-T01) by DECR2 MaxPab polyclonal antibody.

          Lane 1: DECR2 transfected lysate(32.12 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          DECR2 Antibody in Western Blot (WB)
          DECR2 Antibody in Western Blot (WB)
          DECR2 Antibody in Western Blot (WB)
          DECR2 Polyclonal Antibody, MaxPab™

          Product Details

          H00026063-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Rat

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          DECR2 (NP_065715, 1 a.a. approximately 292 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Amount

          50 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MAQPPPDVEG DDCLPAYRHL FCPDLLRDKV AFITGGGSGI GFRIAEIFMR HGCHTVIASR SLPRVLTAAR KLAGATGRRC LPLSMDVRAP PAVMAAVDQA LKEFGRIDIL INCAAGNFLC PAGALSFNAF KTVMDIDTSG TFNVSRVLYE KFFRDHGGVI VNITATLGNR GQALQVHAGS AKAAVDAMTR HLAVEWGPQN IRVNSLAPGP ISGTEGLRRL GGPQASLSTK VTASPLQRLG NKTEIAHSVL YLASPLASYV TGAVLVADGG AWLTFPNGVK GLPDFASFSA KL

          Target Information

          DECR2 (2, 4-dienoyl-CoA reductase 2), also known as PDCR (peroxisomal 2, 4-dienoyl-CoA reductase) or SDR17C1, is a 292 amino acid member of the short-chain dehydrogenases/reductases (SDR) protein family and the 2, 4-dienoyl-CoA reductase protein subfamily. Localized to the peroxisome, DECR2 is an auxiliary enzyme of beta-oxidation that catalyzes the NADP-dependent reduction of 2, 4-dienoyl-CoA to yield trans-3-enoyl-CoA. DECR2 has also been shown to have catalytic activity towards 2, 4, 7, 10, 13, 16, 19-docosaheptaenoyl-CoA and short and medium chain 2, 4-dienoyl-CoAs, suggesting that DECR2 is not a rate limiting step in the degradation of docosahexaenoic acid in the peroxisome. DECR2 is expressed as three isoforms produced by alternative splicing events.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 2,4-dienoyl-CoA reductase 2; 2,4-dienoyl-CoA reductase 2, peroxisomal; 2-4-dienoyl-Coenzyme A reductase 2, peroxisomal; DCR-AKL; pDCR; peroxisomal 2,4-dienoyl-CoA reductase; Peroxisomal 2,4-dienoyl-CoA reductase [(3E)-enoyl-CoA-producing]; putative peroxisomal 2,4-dienoyl-CoA reductase; putative peroxisomal 24-dienoyl-CoA reductase; pVI-AKL; Short chain dehydrogenase/reductase family 17C member 1; unnamed protein product

          View more View less

          Gene Aliases: Dcrakl; DECR2; PDCR; putativeperoxisomal24-dienoyl-CoAreductase; SDR17C1

          View more View less

          UniProt ID: (Human) Q9NUI1, (Rat) Q9Z2M4

          View more View less

          Entrez Gene ID: (Human) 26063, (Rat) 64461

          View more View less

          Function(s)
          protein binding 2,4-dienoyl-CoA reductase (NADPH) activity oxidoreductase activity trans-2-enoyl-CoA reductase (NADPH) activity macromolecular complex binding GTPase binding receptor binding reductase
          Process(es)
          lipid metabolic process fatty acid metabolic process unsaturated fatty acid biosynthetic process fatty acid catabolic process fatty acid beta-oxidation using acyl-CoA oxidase oxidation-reduction process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline