Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • DGAT2 Antibodies

          Invitrogen

          DGAT2 Polyclonal Antibody

          View all (10) DGAT2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite DGAT2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          DGAT2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          DGAT2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          DGAT2 Antibody (PA5-75616) in WB

          Western blot analysis of DGAT2 in Lane 1: C6 whole cell lysate (40 µg), Lane 2: AML-12 whole cell lysate (40 µg), Lane 3: A375 whole cell lysate (40 µg), Lane 4: L02 whole cell lysate (40 µg), Lane 5: HepG2 whole cell lysate (40 µg). Samples were incubated with DGAT2 polyclonal antibody (Product # PA5-75616) at a dilution of 1:500. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          DGAT2 Antibody in Western Blot (WB)

          Product Details

          PA5-75616

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to residues in Human DGAT2.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.2, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2719344

          Product Specific Information

          The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

          Target Information

          Acyl-coenzyme A:diacylglycerol acyltransferase, also known as DGAT1 and ARGP1, is a microsomal enzyme that assists in the synthesis of fatty acids into triglycerides. DGAT1 catalyzes the terminal and only committed step in triacylglycerol synthesis by using diacylglycerol (DAG) and fatty acyl CoA as substrates. DGAT1 plays a fundamental role in the metabolism of cellular diacylglycerol and is important in higher eukaryotes for physiologic processes involving triacylglycerol metabolism, such as intestinal fat absorption, lipoprotein assembly, adipose tissue form-ation and lactation. DGAT2, which has no homology to DGAT1, differs from DGAT1 in that its activity has been shown to be inhibited by MgCl in an in vitro assay. DGAT2 is expressed primarily in liver and white adipose tissue, which suggests that it plays an important role in mammalian triglyceride metabolism.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Acyl-CoA retinol O-fatty-acyltransferase; ARAT; DGAT-2; diacylglycerol acyltransferase 2; Diacylglycerol O-acyltransferase 2; diacylglycerol O-acyltransferase homolog 2; diacylglycerol O-acyltransferase-like protein 2; Diglyceride acyltransferase 2; DKFZp686A15125; GS1999full; retinol O-fatty-acyltransferase

          View more View less

          Gene Aliases: 0610010B06Rik; ARAT; DGAT-2; DGAT2; GS1999FULL; HMFN1045; UNQ738/PRO1433

          View more View less

          UniProt ID: (Human) Q96PD7, (Rat) Q5FVP8, (Mouse) Q9DCV3

          View more View less

          Entrez Gene ID: (Human) 84649, (Rat) 252900, (Mouse) 67800

          View more View less

          Function(s)
          2-acylglycerol O-acyltransferase activity diacylglycerol O-acyltransferase activity protein binding O-acyltransferase activity transferase activity transferase activity, transferring acyl groups protein homodimerization activity retinol O-fatty-acyltransferase activity transferase activity, transferring acyl groups other than amino-acyl groups acyltransferase
          Process(es)
          glycerol metabolic process lipid metabolic process monoacylglycerol biosynthetic process diacylglycerol biosynthetic process response to nutrient positive regulation of triglyceride biosynthetic process triglyceride biosynthetic process lipid storage low-density lipoprotein particle clearance long-chain fatty-acyl-CoA metabolic process cellular triglyceride homeostasis retinol metabolic process cholesterol homeostasis positive regulation of gluconeogenesis negative regulation of fatty acid oxidation diacylglycerol metabolic process regulation of lipoprotein metabolic process fatty acid homeostasis fat pad development cellular response to oleic acid regulation of cholesterol metabolic process regulation of plasma lipoprotein particle levels
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5btrv:80/100.66.77.21:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline